Merck
  • Maternal adverse effects with different loading infusion rates of antenatal magnesium sulphate for preterm fetal neuroprotection: the IRIS randomised trial.
Maternal adverse effects with different loading infusion rates of antenatal magnesium sulphate for preterm fetal neuroprotection: the IRIS randomised trial.
BJOG : an international journal of obstetrics and gynaecology (2014-01-07)
E S Bain, P F Middleton, L N Yelland, P J Ashwood, C A Crowther
ABSTRACT
To evaluate a slower (compared with a standard) infusion rate of the loading dose of magnesium sulphate for preterm fetal neuroprotection as a strategy to reduce maternal adverse effects. Randomised controlled trial. South Australian maternity hospital. Fifty-one women at <30 weeks of gestation, where birth was planned or expected within 24 hours. Women received a loading infusion of 4 g of magnesium sulphate over either 60 or 20 minutes (followed by maintenance of 1 g/hour until birth, or for up to 24 hours). Any maternal adverse effects associated with the infusion. Overall, 71% of women experienced adverse effects during the first hour of their infusion; the difference between groups was not significant [15/25 (60%) 60-minute loading; 21/26 (81%) 20-minute loading; risk ratio (RR) 0.74; 95% confidence interval (95% CI) 0.51-1.08]. Although no serious maternal complications occurred, adverse effects led to three women ceasing the loading treatment (1/25 in the 60-minute loading group; 2/26 in the 20-minute loading group; RR 0.52; 95% CI 0.05-5.38). Women in the 60-minute loading group experienced significantly less warmth and flushing at 20 minutes into the infusion (7/25 in the 60-minute loading group; 15/26 in the 20-minute loading group; RR 0.49; 95% CI 0.24-0.99). No other differences between groups for maternally reported and clinical adverse effects were shown. A slower rate of administering the loading dose of magnesium sulphate did not reduce the occurrence of maternal adverse effects overall. Flushing and warmth at 20 minutes into the infusion was reduced with a slower infusion.
Sigma-Aldrich
Magnesium sulfate heptahydrate, BioXtra, ≥99.0%
Sigma-Aldrich
Magnesium sulfate heptahydrate, BioReagent, for molecular biology, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Magnesium sulfate heptahydrate, BioUltra, ≥99.5% (KT)
Sigma-Aldrich
Magnesium sulfate heptahydrate, ACS reagent, ≥98%
Sigma-Aldrich
Magnesium sulfate heptahydrate, 99.5-100.5% (calc. to the dried substance), meets analytical specification of Ph. Eur., BP,USP, FCC
Sigma-Aldrich
Magnesium sulfate heptahydrate, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Magnesium sulfate heptahydrate, puriss. p.a., ACS reagent, ≥99.0% (KT)
Sigma-Aldrich
Magnesium sulfate solution, BioUltra, for molecular biology
Sigma-Aldrich
Magnesium sulfate, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Magnesium sulfate solution, for molecular biology, 1.00 M±0.04 M
Sigma-Aldrich
Magnesium sulfate, ≥99.99% trace metals basis
Sigma-Aldrich
Magnesium sulfate, puriss. p.a., drying agent, anhydrous, ≥98.0% (KT), powder (very fine)
Sigma-Aldrich
Magnesium sulfate, anhydrous, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Magnesium sulfate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Magnesium sulfate, anhydrous, reagent grade, ≥97%