- Molecular characterization of an invasive phenotype of group A Streptococcus arising during human infection using whole genome sequencing of multiple isolates from the same patient.
Molecular characterization of an invasive phenotype of group A Streptococcus arising during human infection using whole genome sequencing of multiple isolates from the same patient.
The Journal of infectious diseases (2013-12-07)
Anthony R Flores, Pranoti Sahasrabhojane, Miguel Saldaña, Jessica Galloway-Peña, Randall J Olsen, James M Musser, Samuel A Shelburne
PMID24307742
ABSTRACT
Invasive group A streptococcal (GAS) strains often have genetic differences compared to GAS strains from nonsterile sites. Invasive, "hypervirulent" GAS strains can arise from a noninvasive progenitor following subcutaneous inoculation in mice, but such emergence has been rarely characterized in humans. We used whole genome analyses of multiple GAS isolates from the same patient to document the molecular basis for emergence of a GAS strain with an invasive phenotype during human infection. In contrast to previous theories, we found that elimination of production of the cysteine protease SpeB was not necessary for emergence of GAS with an invasive, "hypervirulent" phenotype.
Sigma-Aldrich
Ceftriaxone disodium salt hemi(heptahydrate), third-generation cephalosporin antibiotic