Merck
  • Fluoroscopically guided cervical interlaminar epidural injections using the midline approach: an analysis of epidurography contrast patterns.
Fluoroscopically guided cervical interlaminar epidural injections using the midline approach: an analysis of epidurography contrast patterns.
Anesthesia and analgesia (2009-04-18)
Kwang Su Kim, Sung Sik Shin, Tae Sam Kim, Chang Young Jeong, Myung Ha Yoon, Jeong Il Choi
ABSTRACT
The purpose of this study was to evaluate epidurography contrast patterns in fluoroscopically guided cervical interlaminar epidural injections using the midline approach. All epidural injections were performed at the C6-7 level in the prone position. Epidurography was performed using a fixed amount of iotrolan 240 mg I/mL (1, 2, 3 mL). The rate of ventral epidural spread was 56.7% in Group A (1 mL), 90% in Group B (2 mL), and 93.3% in Group C (3 mL). Two milliliters of contrast solution can provide optimal dispersion of contrast in a ventral and longitudinal spread.
Product Description
Iotrolan, European Pharmacopoeia (EP) Reference Standard
Iotrolan for system suitability, European Pharmacopoeia (EP) Reference Standard