Merck
  • Severe symptomatic aseptic chemical meningitis following myelography: the role of procalcitonin.
Severe symptomatic aseptic chemical meningitis following myelography: the role of procalcitonin.
Neurology (2004-10-13)
Andreas Bender, Matthias Elstner, Robert Paul, Andreas Straube
ABSTRACT
Symptomatic aseptic, chemical meningitis is a rare complication of myelography. Its acute clinical course and standard laboratory findings are indistinguishable from those of bacterial meningitis. The authors present a case of severe postmyelographic chemical meningitis and compare CSF and serum inflammatory markers to a group of seven patients with proven bacterial meningitis. As in viral meningitis, procalcitonin might be able to discriminate between bacterial and chemical causes of CNS inflammation.
Product Description
Iotrolan, European Pharmacopoeia (EP) Reference Standard
Iotrolan for system suitability, European Pharmacopoeia (EP) Reference Standard