Merck
  • Exome sequence reveals mutations in CoA synthase as a cause of neurodegeneration with brain iron accumulation.
Exome sequence reveals mutations in CoA synthase as a cause of neurodegeneration with brain iron accumulation.
American journal of human genetics (2013-12-24)
Sabrina Dusi, Lorella Valletta, Tobias B Haack, Yugo Tsuchiya, Paola Venco, Sebastiano Pasqualato, Paola Goffrini, Marco Tigano, Nikita Demchenko, Thomas Wieland, Thomas Schwarzmayr, Tim M Strom, Federica Invernizzi, Barbara Garavaglia, Allison Gregory, Lynn Sanford, Jeffrey Hamada, Conceição Bettencourt, Henry Houlden, Luisa Chiapparini, Giovanna Zorzi, Manju A Kurian, Nardo Nardocci, Holger Prokisch, Susan Hayflick, Ivan Gout, Valeria Tiranti
ABSTRACT
Neurodegeneration with brain iron accumulation (NBIA) comprises a clinically and genetically heterogeneous group of disorders with progressive extrapyramidal signs and neurological deterioration, characterized by iron accumulation in the basal ganglia. Exome sequencing revealed the presence of recessive missense mutations in COASY, encoding coenzyme A (CoA) synthase in one NBIA-affected subject. A second unrelated individual carrying mutations in COASY was identified by Sanger sequence analysis. CoA synthase is a bifunctional enzyme catalyzing the final steps of CoA biosynthesis by coupling phosphopantetheine with ATP to form dephospho-CoA and its subsequent phosphorylation to generate CoA. We demonstrate alterations in RNA and protein expression levels of CoA synthase, as well as CoA amount, in fibroblasts derived from the two clinical cases and in yeast. This is the second inborn error of coenzyme A biosynthesis to be implicated in NBIA.
Sigma-Aldrich
Iron, ≥99%, reduced, powder (fine)
Sigma-Aldrich
Iron, foil, thickness 0.25 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Iron, wire, diam. 1.0 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Iron, granular, 10-40 mesh, >99.99% trace metals basis
Sigma-Aldrich
Carbonyl iron, ≥97% Fe basis
Iron, IRMM®, certified reference material, 0.5 mm wire
Sigma-Aldrich
Iron, chips, 99.98% trace metals basis
Sigma-Aldrich
Iron, foil, thickness 0.1 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Iron, powder, −325 mesh, 97%
Sigma-Aldrich
Coenzyme A hydrate, ≥85% (UV, HPLC)
Sigma-Aldrich
Iron, puriss. p.a., carbonyl-Iron powder, low in magnesium and manganese compounds, ≥99.5% (RT)