Merck
  • IL-4 directly signals tissue-resident macrophages to proliferate beyond homeostatic levels controlled by CSF-1.
IL-4 directly signals tissue-resident macrophages to proliferate beyond homeostatic levels controlled by CSF-1.
The Journal of experimental medicine (2013-10-09)
Stephen J Jenkins, Dominik Ruckerl, Graham D Thomas, James P Hewitson, Sheelagh Duncan, Frank Brombacher, Rick M Maizels, David A Hume, Judith E Allen
ABSTRACT
Macrophages (MΦs) colonize tissues during inflammation in two distinct ways: recruitment of monocyte precursors and proliferation of resident cells. We recently revealed a major role for IL-4 in the proliferative expansion of resident MΦs during a Th2-biased tissue nematode infection. We now show that proliferation of MΦs during intestinal as well as tissue nematode infection is restricted to sites of IL-4 production and requires MΦ-intrinsic IL-4R signaling. However, both IL-4Rα-dependent and -independent mechanisms contributed to MΦ proliferation during nematode infections. IL-4R-independent proliferation was controlled by a rise in local CSF-1 levels, but IL-4Rα expression conferred a competitive advantage with higher and more sustained proliferation and increased accumulation of IL-4Rα(+) compared with IL-4Rα(-) cells. Mechanistically, this occurred by conversion of IL-4Rα(+) MΦs from a CSF-1-dependent to -independent program of proliferation. Thus, IL-4 increases the relative density of tissue MΦs by overcoming the constraints mediated by the availability of CSF-1. Finally, although both elevated CSF1R and IL-4Rα signaling triggered proliferation above homeostatic levels, only CSF-1 led to the recruitment of monocytes and neutrophils. Thus, the IL-4 pathway of proliferation may have developed as an alternative to CSF-1 to increase resident MΦ numbers without coincident monocyte recruitment.
Sigma-Aldrich
Macrophage Colony-Stimulating Factor from mouse, M-CSF, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture