Merck
  • Drug rescue distinguishes between different structural models of human P-glycoprotein.
Drug rescue distinguishes between different structural models of human P-glycoprotein.
Biochemistry (2013-10-03)
Tip W Loo, David M Clarke
ABSTRACT
There is no high-resolution crystal structure of the human P-glycoprotein (P-gp) drug pump. Homology models of human P-gp based on the crystal structures of mouse or Caenorhabditis elegans P-gps show large differences in the orientation of transmembrane segment 5 (TM5). TM5 is one of the most important transmembrane segments involved in drug-substrate interactions. Drug rescue of P-gp processing mutants containing an arginine at each position in TM5 was used to identify positions facing the lipid or internal aqueous chamber. Only the model based on the C. elegans P-gp structure was compatible with the drug rescue results.
Sigma-Aldrich
Cyclosporin A, BioReagent, from Tolypocladium inflatum, for molecular biology, ≥95%
Sigma-Aldrich
Cyclosporin A, 97.0-101.5% (on dried basis)
Sigma-Aldrich
Cyclosporin A, from Tolypocladium inflatum, ≥95% (HPLC), solid
Supelco
Cyclosporin A, VETRANAL®, analytical standard
Supelco
Cyclosporin A solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Cyclosporine, Pharmaceutical Secondary Standard; Certified Reference Material