Merck
  • The dependence receptor TrkC triggers mitochondria-dependent apoptosis upon Cobra-1 recruitment.
The dependence receptor TrkC triggers mitochondria-dependent apoptosis upon Cobra-1 recruitment.
Molecular cell (2013-09-17)
Gabriel Ichim, Anne-Laure Genevois, Marie Ménard, Li-Ying Yu, Juliana M Coelho-Aguiar, Fabien Llambi, Loraine Jarrosson-Wuilleme, Jonathan Lefebvre, David Tulasne, Elisabeth Dupin, Nicole Le Douarin, Urmas Arumäe, Servane Tauszig-Delamasure, Patrick Mehlen
ABSTRACT
The neurotrophin receptor TrkC was recently identified as a dependence receptor, and, as such, it triggers apoptosis in the absence of its ligand, NT-3. The molecular mechanism for apoptotic engagement involves the double cleavage of the receptor's intracellular domain, leading to the formation of a proapoptotic "killer" fragment (TrkC KF). Here, we show that TrkC KF interacts with Cobra1, a putative cofactor of BRCA1, and that Cobra1 is required for TrkC-induced apoptosis. We also show that, in the developing chick neural tube, NT-3 silencing is associated with neuroepithelial cell death that is rescued by Cobra1 silencing. Cobra1 shuttles TrkC KF to the mitochondria, where it promotes Bax activation, cytochrome c release, and apoptosome-dependent apoptosis. Thus, we propose that, in the absence of NT-3, the proteolytic cleavage of TrkC leads to the release of a killer fragment that triggers mitochondria-dependent apoptosis via the recruitment of Cobra1.
Sigma-Aldrich
ProteoMass Cytochrome c MALDI-MS Standard, vial of 10 nmol, (M+H+) 12,361.96 Da by calculation
Sigma-Aldrich
Cytochrome c from Saccharomyces cerevisiae, ≥85% based on Mol. Wt. 12,588 basis
Sigma-Aldrich
Cytochrome c from equine heart, BioUltra, ≥99% (SDS-PAGE), powder, suitable for mammalian cell culture
Sigma-Aldrich
Cytochrome c from bovine heart, ≥95% (GE)
Sigma-Aldrich
Cytochrome c from bovine heart, ≥95% based on Mol. Wt. 12,327 basis, powder, suitable for mammalian cell culture
Sigma-Aldrich
Cytochrome c from equine heart, ≥95% based on Mol. Wt. 12,384 basis
Sigma-Aldrich
Cytochrome c from equine heart, ≥95% (SDS-PAGE)
Sigma-Aldrich
Cytochrome c from equine heart, BioReagent, suitable for GFC marker
Sigma-Aldrich
Cytochrome c from bovine heart, ≥95% based on Mol. Wt. 12,327 basis
Sigma-Aldrich
Cytochrome c from pigeon breast muscle, ≥95% based on Mol. Wt. 12,173 basis