Merck
  • Dimethyl fumarate regulates histone deacetylase expression in astrocytes.
Dimethyl fumarate regulates histone deacetylase expression in astrocytes.
Journal of neuroimmunology (2013-08-07)
Sergey Kalinin, Paul E Polak, Shao Xia Lin, David Braun, Marina Guizzetti, Xiaolu Zhang, Israel Rubinstein, Douglas L Feinstein
ABSTRACT
We previously showed that dimethyl fumarate (DMF) reduces inflammatory activation in astrocytes, involving activation of transcription factor Nrf2. However, the pathways causing Nrf2 activation were not examined. We now show that DMF modifies expression of histone deacetylases (HDACs) in primary rat astrocytes. After 4h incubation, levels of HDAC1, 2, and 4 mRNAs were increased by DMF; however, after 24h, levels returned to or were below control values. At that time, HDAC protein levels and overall activity were also reduced by DMF. Stimulation of astrocytes with pro-inflammatory cytokines significantly increased HDAC mRNA levels after 24h, although protein levels were not increased at that time point. In the presence of cytokines, DMF reduced HDAC mRNAs, proteins, and activity. Proteomic analysis of DMF-treated astrocytes identified 8 proteins in which lysine acetylation was increased by DMF, including histones H2a.1 and H3.3. A role for HDACs in mediating DMF actions is suggested by findings that the selective HDAC inhibitor SAHA increased nuclear Nrf2:DNA binding activity, reduced inflammatory activation of astrocytes which was reversed by a selective inhibitor of the Nrf2 target gene heme-oxygenase 1. These data show that DMF regulates astrocyte HDAC expression, which could contribute to Nrf2 activation, suppression of inflammatory responses and cause long-lasting changes in gene expression.
Sigma-Aldrich
Dimethyl fumarate, 97%