Merck
A versatile light microscope heating stage for biological temperatures.
Journal of microscopy (1986-06-01)
R H Lowndes, M B Hallett
ABSTRACT
A light microscope heating stage suitable for biological temperatures is described. A novel feature of the design is the use of cupro-nickel foil heaters (0.1 mm thick). With a temperature sensor mounted on the slide clip and a temperature controller unit, this apparatus maintains a temperature of 310 K with a precision of 0.1 K. The apparatus is inexpensive, reliable, not confined to use with one microscope and does not interfere significantly with the stage movement or optical alignment.
Product Description
Nickel, foil, 6mm disks, thickness 0.02mm, 99.9%
Nickel, foil, 8mm disks, thickness 0.125mm, annealed, 99%
Nickel, foil, 8mm disks, thickness 0.0125mm, 99.99+%
Nickel, foil, 8mm disks, thickness 0.05mm, as rolled, 100%
Nickel, foil, 8mm disks, thickness 0.03mm, 99.9%
Nickel, foil, 6mm disks, thickness 0.0125mm, 99.9%
Nickel, foil, 100x100mm, thickness 0.125mm, as rolled, 99.999%
Nickel, foil, 6mm disks, thickness 0.038mm, 99.9%
Nickel, foil, 8mm disks, thickness 0.05mm, 99.9%
Nickel, foil, 100x100mm, thickness 0.25mm, as rolled, 99.99+%
Nickel, foil, 6mm disks, thickness 0.3mm, as rolled, 100%
Nickel, foil, 6mm disks, thickness 0.0125mm, annealed, 99%
Nickel, foil, 100x100mm, thickness 1.0mm, annealed, 99.98%
Nickel, foil, 6mm disks, thickness 0.15mm, annealed, 99%
Nickel, foil, 8mm disks, thickness 0.01mm, 99.95%
Nickel, foil, 8mm disks, thickness 0.125mm, annealed, 99.98%
Nickel, foil, 6mm disks, thickness 0.3mm, annealed, 99%
Nickel, foil, not light tested, 100x100mm, thickness 0.002mm, 99.95%
Nickel, foil, 0.5m coil, thickness 0.075mm, as rolled, 99.98%
Nickel, foil, 4mm disks, thickness 0.025mm, 99.9%
Nickel, foil, 0.5m coil, thickness 0.004mm, hard, 99.98%
Nickel, foil, 4mm disks, thickness 0.025mm, as rolled, 100%
Nickel, foil, 2m coil, thickness 0.15mm, annealed, 99%
Nickel, foil, 0.5m coil, thickness 0.025mm, coil width 600mm, as rolled, 99%
Nickel, foil, 10m coil, thickness 0.125mm, coil width 50mm, annealed, 99%
Nickel, foil, 0.5m coil, thickness 0.125mm, annealed, 99%
Nickel, foil, 10mm disks, thickness 0.005mm, as rolled, 99.9%
Nickel, foil, 2m coil, thickness 0.5mm, annealed, 99.98%
Nickel, foil, 0.5m coil, thickness 0.038mm, 99.9%
Nickel, foil, 4mm disks, thickness 0.02mm, 99.9%