Merck
Scaling limitations of laser-fabricated nerve electrode arrays.
Conference proceedings : ... Annual International Conference of the IEEE Engineering in Medicine and Biology Society. IEEE Engineering in Medicine and Biology Society. Annual Conference (2009-01-24)
Christian Henle, Martin Schuettler, Juan S Ordonez, Thomas Stieglitz
ABSTRACT
A laser technology for manufacturing implantable electrode arrays, using spin-on polydimethylsiloxane (PDMS) and platinum foil as materials, was investigated in respect to its scaling limitations. The following aspects were analyzed: Minimal width and centre-to-centre distance of platinum tracks, the ability of spin-on PDMS to flow between platinum tracks with very narrow gaps and the electrical insulation properties of thin spin-coated PDMS.
Product Description
Platinum, foil, 0.2m coil, thickness 0.025mm, coil width 1.6mm, as rolled, 99.95%
Platinum, foil, 0.5m coil, thickness 0.025mm, coil width .4mm, as rolled, 99.95%
Platinum, foil, 25x25mm, thickness 0.125mm, as rolled, 99.95%
Platinum, foil, 50x50mm, thickness 0.125mm, as rolled, 99.99+%
Platinum, foil, 0.1m coil, thickness 0.15mm, coil width .9mm, as rolled, 99.99%
Platinum, foil, 0.05m coil, thickness 0.029mm, coil width .34mm, annealed, 99.99%
Platinum, foil, 0.1m coil, thickness 0.025mm, coil width 1.6mm, as rolled, 99.95%
Platinum, foil, 50x50mm, thickness 0.1mm, as rolled, 99.99+%
Platinum, foil, 0.05m coil, thickness 0.025mm, coil width .4mm, as rolled, 99.95%
Platinum, foil, 0.1m coil, thickness 0.029mm, coil width .34mm, annealed, 99.99%
Platinum, foil, 0.5m coil, thickness 0.045mm, coil width .75mm, as rolled, 99.95%
Platinum, foil, not light tested, 50x50mm, thickness 0.01mm, as rolled, 99.95%
Platinum, foil, 30x30mm, thickness 0.2mm, as rolled, 99.95%
Platinum, foil, 15x15mm, thickness 0.2mm, as rolled, 99.95%
Platinum, foil, 25x25mm, thickness 0.125mm, as rolled, 99.99+%
Platinum, foil, not light tested, 50x50mm, thickness 0.0025mm, as rolled, 99.95%
Platinum, foil, not light tested, 50x50mm, thickness 0.005mm, as rolled, 99.95%
Platinum, foil, not light tested, 25x25mm, thickness 0.0075mm, as rolled, 99.95%
Platinum, foil, light tested, 50x50mm, thickness 0.05mm, as rolled, 99.95%
Platinum, foil, not light tested, 25x25mm, thickness 0.00075mm, permanent polyester support, 99.85%
Platinum, foil, light tested, 50x50mm, thickness 0.005mm, as rolled, 99.95%
Platinum, foil, not light tested, 100x100mm, thickness 0.003mm, as rolled, 99.95%
Platinum, foil, not light tested, 25x25mm, thickness 0.004mm, as rolled, 99.95%
Platinum, foil, 0.5m coil, thickness 0.025mm, coil width .85mm, as rolled, 99.95%
Platinum, foil, 0.2m coil, thickness 0.025mm, coil width 5mm, as rolled, 99.95%
Platinum, foil, 0.2m coil, thickness 0.029mm, coil width .34mm, annealed, 99.99%
Platinum, foil, 0.5m coil, thickness 0.025mm, coil width 5mm, as rolled, 99.95%
Platinum, foil, light tested, 25x25mm, thickness 0.025mm, as rolled, 99.99+%
Platinum, foil, 0.2m coil, thickness 0.045mm, coil width .75mm, as rolled, 99.95%
Platinum, foil, not light tested, 50x50mm, thickness 0.00075mm, permanent polyester support, 99.85%