- Surface-enhanced Raman spectrometry of triamterene on a silver substrate prepared by the nitric acid etching method.
Surface-enhanced Raman spectrometry of triamterene on a silver substrate prepared by the nitric acid etching method.
Talanta (1997-02-01)
A Rupérez, J J Laserna
PMID18966737
ABSTRACT
Surface-enhanced Raman scattering (SERS) spectrometry of the diuretic drug triamterene is discussed. The SERS-active substrate used is a silver foil etched with nitric acid. The influence of solvent and sample doping method on sensivity, intercept and shape of the calibration graphs is discussed.
Product Description
Silver, foil, 0.2m coil, thickness 0.01mm, 99.95+%
Silver, foil, 25x25mm, thickness 0.1mm, as rolled, 99.95+%
Silver, foil, not light tested, 100x100mm, thickness 0.0125mm, 99.95+%
Silver, foil, not light tested, 300x300mm, thickness 0.025mm, as rolled, 99.95+%
Silver, foil, light tested, 100x100mm, thickness 0.025mm, annealed, 99.95+%
Silver, foil, light tested, 150x150mm, thickness 0.035mm, as rolled, 99.95+%
Silver, foil, not light tested, 150x150mm, thickness 0.01mm, 99.95+%
Silver, foil, light tested, 150x150mm, thickness 0.05mm, annealed, 99.95+%
Silver, foil, not light tested, 100x100mm, thickness 0.025mm, annealed, 99.95+%
Silver, foil, not light tested, 25x25mm, thickness 0.005mm, 99.97%
Silver, foil, not light tested, 150x150mm, thickness 0.025mm, as rolled, 99.95+%
Silver, foil, not light tested, 25x25mm, thickness 0.006mm, 99.97%
Silver, foil, not light tested, 150x150mm, thickness 0.02mm, 99.97%
Silver, foil, light tested, 300x300mm, thickness 0.035mm, as rolled, 99.95+%
Silver, foil, light tested, 25x25mm, thickness 0.015mm, 99.97%
Silver, foil, not light tested, 250x300mm, thickness 0.025mm, annealed, 99.95+%
Silver, foil, light tested, 100x100mm, thickness 0.05mm, as rolled, 99.95+%
Silver, foil, not light tested, 100x100mm, thickness 0.006mm, 99.97%
Silver, foil, light tested, 25x25mm, thickness 0.05mm, as rolled, 99.95+%
Silver, foil, light tested, 100x100mm, thickness 0.025mm, as rolled, 99.99+%
Silver, foil, light tested, 50x50mm, thickness 0.02mm, 99.97%
Silver, foil, light tested, 50x50mm, thickness 0.015mm, 99.97%
Silver, foil, not light tested, 50x50mm, thickness 0.008mm, 99.95+%
Silver, foil, 10m coil, thickness 0.0125mm, coil width 9mm, 99.95+%
Silver, foil, 0.5m coil, thickness 0.035mm, as rolled, 99.95+%
Silver, foil, 50x50mm, thickness 0.125mm, as rolled, 99.99+%
Silver, foil, 0.5m coil, thickness 0.008mm, 99.95+%
Silver, foil, 50x50mm, thickness 0.1mm, annealed, 99.95+%
Silver, foil, 0.5m coil, thickness 0.003mm, 99.9%
Silver, foil, 150x150mm, thickness 0.1mm, annealed, 99.95+%