Merck
A flattening filter for brachytherapy skin irradiation.
Physics in medicine and biology (2002-04-05)
Tomas Kron, Mamoon Haque, Kristie Foulke, Robert Jeraj
ABSTRACT
Radioactive sources in close contact offer an alternative to superficial radiation in the treatment of skin lesions. A flattening filter was designed for a lead surface applicator to improve the skin dose distribution of a high dose rate (HDR) brachytherapy unit (Nucletron). At three heights from the opening (10, 15 and 25 mm) of the cylindrical applicator, the 192Ir source can be driven into the centre of the applicator. Thin sheets of lead foil (0.2 mm) were cut into circular shapes and placed in the opening to build a cylindrical cone that acts as a flattening filter. The shape of the cone was optimized in an iterative process using a spreadsheet and the resulting dose distribution under the applicator was determined using radiosensitive film. The use of the filter improved the dose distribution in a plane perpendicular to the beam axis to be within +/- 5% of the central axis dose. The present applicator and flattening filter together with an HDR brachytherapy unit offer an alternative for skin irradiation where a superficial unit is not available or will be replaced with a more flexible device. As the depth dose characteristics can be modified using different source-to-surface distances, the dose throughout the patient's skin can be shaped as desired by the radiation oncologist using a compensator design type approach.
Product Description
Lead, foil, not light tested, 25x25mm, thickness 0.0125mm, 99.99+%
Lead, foil, light tested, 50x50mm, thickness 0.05mm, as rolled, 97% (Sn 1.5%, Sb 1.5%)
Lead, foil, 2m coil, thickness 1.0mm, as rolled, 99.95%
Lead, foil, 4mm disks, thickness 0.0125mm, 99.99+%
Lead, foil, 25mm disks, thickness 0.25mm, as rolled, 99.95%
Lead, foil, 25mm disks, thickness 0.1mm, as rolled, 99.95%
Lead, foil, 2m coil, thickness 0.15mm, as rolled, 99.95%
Lead, foil, 2m coil, thickness 0.12mm, coil width 25.4mm, as rolled, 99.95%
Lead, foil, 100x100mm, thickness 1.0mm, as rolled, 99.95%
Lead, foil, 10m coil, thickness 0.1mm, as rolled, 99.95%
Lead, foil, 10mm disks, thickness 0.55mm, as rolled, 100%
Lead, foil, 150x150mm, thickness 0.25mm, as rolled, 99.95%
Lead, foil, 15mm disks, thickness 0.05mm, 99.99+%
Lead, foil, 5m coil, thickness 0.05mm, as rolled, 97% (sn 1.5%, sb 1.5%)
Lead, foil, 10mm disks, thickness 0.25mm, as rolled, 99.99+%
Lead, foil, 10mm disks, thickness 0.015mm, 99.99+%
Lead, foil, 25mm disks, thickness 0.05mm, 99.99+%
Lead, foil, 10mm disks, thickness 0.04mm, 99.99+%
Lead, foil, 4mm disks, thickness 0.05mm, 99.99+%
Lead, foil, 15mm disks, thickness 0.125mm, as rolled, 99.95%
Lead, foil, 1m coil, thickness 1.0mm, as rolled, 99.95%
Lead, foil, 25x25mm, thickness 0.5mm, as rolled, 99.95%
Lead, foil, 4mm disks, thickness 0.25mm, as rolled, 99.95%
Lead, foil, 10mm disks, thickness 0.1mm, as rolled, 99.95%
Lead, foil, 15mm disks, thickness 0.1mm, as rolled, 99.95%
Lead, foil, 50x50mm, thickness 1.5mm, as rolled, 99.99+%
Lead, foil, 50mm disks, thickness 0.15mm, as rolled, 99.95%
Lead, foil, 1m coil, thickness 0.016mm, as rolled, 96% (sn 1.5%, sb 2.5%)
Lead, foil, 15mm disks, thickness 0.009mm, 99.99+%
Lead, foil, 5m coil, thickness 0.1mm, as rolled, 99.95%