Merck
  • Improved method for the storage of groundwater samples containing volatile organic analytes
Improved method for the storage of groundwater samples containing volatile organic analytes
Archives of environmental contamination and toxicology (1999-02-27)
Kovacs, Kampbell
ABSTRACT
The sorption of volatile organic analytes from water samples by the Teflon septum surface used with standard glass 40-ml sample collection vials was investigated. Analytes tested included alkanes, isoalkanes, olefins, cycloalkanes, a cycloalkene, monoaromatics, a polynuclear aromatic, and two chloroethenes. Both laboratory prepared test mix solutions and petroleum contaminated groundwater from three field sites were tested. A rapid loss of n-alkane and isoalkane concentrations (>10%) was observed within 24 h when stored at room temperature. Aliphatic losses were also observed (>10%) over a 21-day holding period when samples were held at 4 degrees C. Loss of the less sorptive analytes was demonstrated by exposing analyte solutions to greater Teflon surface areas. The demonstrated sorption of aliphatics from water samples by Teflon-lined septa indicates that the accuracy of volatile petroleum hydrocarbon determinations may be reduced by the traditional storage method. An alternative storage protocol is reported combining a lead foil septum surface and 1% (w/w) tribasic sodium phosphate dodecahydrate (Na3PO4. 12H2O) preservative. This method prevented loss of the test analytes, including alkanes and isoalkanes for at least 21 days at room temperature.
Product Description
Lead, foil, not light tested, 50x50mm, thickness 0.01mm, 99.99+%
Lead, foil, not light tested, 25x25mm, thickness 0.008mm, 99.99+%
Lead, foil, not light tested, 150x150mm, thickness 0.05mm, 99.99+%
Lead, foil, not light tested, 50x50mm, thickness 0.0125mm, 99.99+%
Lead, foil, not light tested, 300x300mm, thickness 0.05mm, as rolled, 97% (Sn 1.5%, Sb 1.5%)
Lead, foil, not light tested, 100x100mm, thickness 0.007mm, 99.99+%
Lead, foil, light tested, 50x50mm, thickness 0.02mm, 99.99+%
Lead, foil, light tested, 25x25mm, thickness 0.009mm, 99.99+%
Lead, foil, light tested, 50x50mm, thickness 0.03mm, 99.99+%
Lead, foil, light tested, 25x25mm, thickness 0.025mm, as rolled, 97% (Sn 1.5%, Sb 1.5%)
Lead, foil, not light tested, 100x100mm, thickness 0.009mm, 99.99+%
Lead, foil, not light tested, 150x150mm, thickness 0.03mm, 99.99+%
Lead, foil, light tested, 25x25mm, thickness 0.015mm, 99.99+%
Lead, foil, light tested, 25x25mm, thickness 0.016mm, as rolled, 96% (Sn 1.5%, Sb 2.5%)
Lead, foil, not light tested, 100x100mm, thickness 0.04mm, 99.99+%
Lead, foil, not light tested, 100x100mm, thickness 0.0125mm, 99.99+%
Lead, foil, light tested, 25x25mm, thickness 0.05mm, as rolled, 97% (Sn 1.5%, Sb 1.5%)
Lead, foil, light tested, 25x25mm, thickness 0.03mm, 99.99+%
Lead, foil, not light tested, 50x50mm, thickness 0.03mm, 99.99+%
Lead, foil, light tested, 25x25mm, thickness 0.05mm, as rolled, 99.99+%
Lead, foil, light tested, 50x50mm, thickness 0.025mm, as rolled, 97% (Sn 1.5%, Sb 1.5%)
Lead, foil, not light tested, 50x50mm, thickness 0.015mm, 99.99+%
Lead, foil, 25mm disks, thickness 0.15mm, as rolled, 99.95%
Lead, foil, 4mm disks, thickness 0.025mm, 99.99+%
Lead, foil, 4mm disks, thickness 0.009mm, 99.99+%
Lead, foil, 2m coil, thickness 0.1mm, as rolled, 99.95%
Lead, foil, 4mm disks, thickness 0.015mm, 99.99+%
Lead, foil, 4mm disks, thickness 0.02mm, 99.99+%
Lead, foil, 25x25mm, thickness 1.0mm, as rolled, 99.5%
Lead, foil, 8mm disks, thickness 0.03mm, 99.99+%