Merck
  • Demonstration of the existence of a second, non-lysosomal glucocerebrosidase that is not deficient in Gaucher disease.
Demonstration of the existence of a second, non-lysosomal glucocerebrosidase that is not deficient in Gaucher disease.
Biochimica et biophysica acta (1993-03-24)
S van Weely, M Brandsma, A Strijland, J M Tager, J M Aerts
ABSTRACT
In addition to the lysosomal glucocerebrosidase, a distinct beta-glucosidase that is also active towards glucosylceramide could be demonstrated in various human tissues and cell types. Subcellular fractionation analysis revealed that the hitherto undescribed glucocerebrosidase is not located in lysosomes but in compartments with a considerably lower density. The non-lysosomal glucocerebrosidase differed in several respects from lysosomal glucocerebrosidase. The non-lysosomal isoenzyme proved to be tightly membrane-bound, whereas lysosomal glucocerebrosidase is weakly membrane-associated. The pH optimum of the non-lysosomal isoenzyme is less acidic than that of lysosomal glucocerebrosidase. Non-lysosomal glucocerebrosidase, in contrast to the lysosomal isoenzyme, was not inhibited by low concentrations of conduritol B-epoxide, was markedly inhibited by taurocholate, was not stimulated in activity by the lysosomal activator protein saposin C, and was not deficient in patients with Gaucher disease. Non-lysosomal glucocerebrosidase proved to be less sensitive to inhibition by castanospermine or deoxynojirimycin but more sensitive to inhibition by D-gluconolactone than the lysosomal glucocerebrosidase. The physiological function of this second, non-lysosomal, glucocerebrosidase is as yet unknown.
Sigma-Aldrich
D-(+)-Gluconic acid δ-lactone, ≥99.0% (GC)
Sigma-Aldrich
Gluconolactone, 99.0-101.0%, meets USP testing specifications