Merck
  • Glycosphingolipid accumulation inhibits cholesterol efflux via the ABCA1/apolipoprotein A-I pathway: 1-phenyl-2-decanoylamino-3-morpholino-1-propanol is a novel cholesterol efflux accelerator.
Glycosphingolipid accumulation inhibits cholesterol efflux via the ABCA1/apolipoprotein A-I pathway: 1-phenyl-2-decanoylamino-3-morpholino-1-propanol is a novel cholesterol efflux accelerator.
The Journal of biological chemistry (2005-05-14)
Elias N Glaros, Woojin Scott Kim, Carmel M Quinn, Jenny Wong, Ingrid Gelissen, Wendy Jessup, Brett Garner
ABSTRACT
Cellular glycosphingolipid (GSL) storage is known to promote cholesterol accumulation. Although physical interactions between GSLs and cholesterol are thought to cause intracellular cholesterol "trapping," it is not known whether cholesterol homeostatic mechanisms are also impaired under these conditions. ApoA-I-mediated cholesterol efflux via ABCA1 (ATP-binding cassette transporter A1) is a key regulator of cellular cholesterol balance. Here, we show that apoA-I-mediated cholesterol efflux was inhibited (by up to 53% over 8 h) when fibroblasts were treated with lactosylceramide or the glucocerebrosidase inhibitor conduritol B epoxide. Furthermore, apoA-I-mediated cholesterol efflux from fibroblasts derived from patients with genetic GSL storage diseases (Fabry disease, Sandhoff disease, and GM1 gangliosidosis) was impaired compared with control cells. Conversely, apoA-I-mediated cholesterol efflux from fibroblasts and cholesterol-loaded macrophage foam cells was dose-dependently stimulated (by up to 6-fold over 8 h) by the GSL synthesis inhibitor 1-phenyl-2-decanoylamino-3-morpholino-1-propanol (PDMP). Unexpectedly, a structurally unrelated GSL synthesis inhibitor, N-butyldeoxynojirimycin, was unable to stimulate apoA-I-mediated cholesterol efflux despite achieving similar GSL depletion. PDMP was found to up-regulate ABCA1 mRNA and protein expression, thereby identifying a contributing mechanism for the observed acceleration of cholesterol efflux to apoA-I. This study reveals a novel defect in cellular cholesterol homeostasis induced by GSL storage and identifies PDMP as a new agent for enhancing cholesterol efflux via the ABCA1/apoA-I pathway.
Sigma-Aldrich
1-Propanol, ≥99%, FG
Sigma-Aldrich
1-Propanol, natural, ≥98%, FG
Supelco
1-Propanol, analytical standard
Sigma-Aldrich
1-Propanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
1-Propanol, ≥99% (GC), purum
Sigma-Aldrich
1-Propanol, ACS reagent, ≥99.5%