Merck
Free radical scavengers and antioxidants from Baccharis grisebachii.
Journal of ethnopharmacology (2004-10-28)
Alejandro Tapia, Jaime Rodriguez, Cristina Theoduloz, Susana Lopez, Gabriela Egly Feresin, Guillermo Schmeda-Hirschmann
ABSTRACT
The exudate and seriated extracts from the aerial parts of Baccharis grisebachii (Asteraceae) which is recommended as a digestive and to relieve gastric ulcers in Argentina, showed activity as free radical scavengers and inhibited lipoperoxidation in erythrocytes. Assay-guided isolation led to seven p-coumaric acid derivatives and six flavonoids as the main active constituents of the crude drug. The activity towards the superoxide anion was mainly due to the flavonoid constituents. 5,7,4'-Trihydroxy-6-methoxyflavone and quercetin presented high activity (64 and 79%) even at 12.5 microg/ml. The xanthine oxidase inhibitory effect of the extracts can be related with the p-coumaric acid derivatives drupanin, 4-acetyl-3,5-diprenylcinnamic acid and trans-ferulic acid O-hexan-3-onyl-ether which showed IC(50) values in the range 28-40 microg/ml. Both p-coumaric acid derivatives and flavonoids inhibited lipoperoxidation in erythrocytes. The highest activity was found for the p-coumaric acid derivatives 4-acetyl-3-prenyl-ethoxycinnamate, 3-prenyl-4-(4'-hydroxydihydrocinnamoyloxy)-cinnamate and trans-ferulic acid O-hexan-3-onyl-ether (69-82%) and the flavonoids 5,7,4'-trihydroxy-6-methoxyflavone, quercetin, 5,7,4'-trihydroxy-6,3'-dimethoxyflavone and 5,7,4'-trihydroxy-6,8-dimethoxyflavone (64-84%) at 100 microg/ml. The most active free radical scavengers measured by the DPPH decoloration assay were the p-coumaric acid derivatives drupanin and trans-ferulic acid O-hexan-3-onyl-ether (27-35% at 10 microg/ml) and the flavonoid quercetin (97 and 23% at 10 and 1 microg/ml, respectively). The results support the use of Baccharis grisebachii in Argentinian traditional medicine.
Product Description
Supelco
trans-Ferulic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
trans-Ferulic acid, matrix substance for MALDI-MS, ≥99.0% (HPLC)
Sigma-Aldrich
trans-Ferulic acid, ≥99%
Sigma-Aldrich
trans-Ferulic acid, 99%