Sigma-Aldrich
Tungsten, foil, thickness 0.25 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Tungsten, powder, 12 μm, 99.9% trace metals basis
Sigma-Aldrich
Molybdenum, powder, <150 μm, 99.99% trace metals basis
Sigma-Aldrich
Molybdenum, foil, thickness 0.1 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Molybdenum, foil, thickness 1.0 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Molybdenum, powder, 10 μm, ≥99.95% trace metals basis
Sigma-Aldrich
Molybdenum, nanopowder, <100 nm particle size (TEM), 99.8% trace metals basis
Sigma-Aldrich
Molybdenum, powder, <150 μm, 99.9% trace metals basis
Sigma-Aldrich
Molybdenum, foil, thickness 0.025 mm, ≥99.9% trace metals basis
Tungsten, wire, straight, 300mm, diameter 0.75mm, 99.9+%
Tungsten, wire, straight, 100mm, diameter 0.125mm, 99.9+%
Tungsten, wire reel, 5m, diameter 0.125mm, clean, 99.95%
Tungsten, wire, straight, 300mm, diameter 0.20mm, 99.9+%
Tungsten, wire, straight, 196mm, diameter 0.30mm, 99.9+%
Tungsten, wire reel, 5m, diameter 0.1mm, clean, 99.95%
Tungsten, wire, straight, 300mm, diameter 0.15mm, 99.9+%
Tungsten, foil, 0.2m coil, thickness 0.04mm, coil width 0.7mm, as rolled, 99.95%
Tungsten, foil, 100x100mm, thickness 0.2mm, as rolled, 99.95%
Tungsten, foil, 0.2m coil, thickness 0.05mm, coil width .51mm, as rolled, 99.95%
Tungsten, foil, 0.2m coil, thickness 0.05mm, coil width 1.5mm, as rolled, 99.95%
Tungsten, foil, 100x100mm, thickness 0.125mm, as rolled, 99.95%
Tungsten, foil, 200x200mm, thickness 0.15mm, as rolled, 99.95%
Tungsten, foil, 0.1m coil, thickness 0.076mm, coil width 2mm, as rolled, 99.95%
Tungsten, foil, 50x50mm, thickness 0.2mm, as rolled, 99.95%
Tungsten, foil, 50x50mm, thickness 0.125mm, as rolled, 99.95%
Tungsten, foil, 0.2m coil, thickness 0.02mm, coil width .1mm, as rolled, 99.95%
Tungsten, foil, 200x200mm, thickness 0.3mm, as rolled, 99.95%
Tungsten, foil, 0.5m coil, thickness 0.025mm, coil width .7mm, as rolled, 99.95%
Tungsten, foil, 150x150mm, thickness 0.5mm, as rolled, 99.95%
Tungsten, foil, 100x100mm, thickness 0.1mm, as rolled, 99.95%