- PRODUCTION OF HYDROGEN SULFIDE BY STREPTOMYCES ODORIFER.
PRODUCTION OF HYDROGEN SULFIDE BY STREPTOMYCES ODORIFER.
Applied microbiology (1964-07-01)
R P COLLINS, H D GAINES
PMID14199024
ABSTRACT
By use of various trapping systems, as well as lead acetate papers, Streptomyces odorifer was shown to produce hydrogen sulfide. Other sulfur-containing compounds may be produced by S. odorifer, but the amounts obtained were too small for detailed analysis. It was suggested that hydrogen sulfide might be a part of the earthy-odor complex produced by S. odorifer.