Merck
  • Reversal of acquired resistance to doxorubicin in P388 murine leukemia cells by tamoxifen and other triparanol analogues.
Reversal of acquired resistance to doxorubicin in P388 murine leukemia cells by tamoxifen and other triparanol analogues.
Cancer research (1984-10-01)
A Ramu, D Glaubiger, Z Fuks
ABSTRACT
The effects of the triparanol analogues chlorotrianisene, clomiphene, tamoxifen, 5-[p-(fluoren-9-ylidenemethyl)phenyl]-2-piperidineethanol (MDL 10393), MDL 8917v, nafoxidine, 2-[p-(6-methoxy-2-phenylinden-3-yl)phenoxy]triethylamine (U-11555A), 2-[p-(3,4-dihydro-6-methoxy-2-phenyl-1-naphthyl)phenoxy]triethylamine (U-10520A), and nitromifene, as well as triparanol itself, were studied in the P388 murine leukemia cell line and in a doxorubicin-resistant subline (P388/ADR). At noninhibitory concentrations, all the analogues increased the sensitivity of P388/ADR cells to doxorubicin but did not have such an effect on the doxorubicin-sensitive cells. Diethylstilbestrol, deacetylated cyclofenil (F6060), hexestrol, and 17 beta-estradiol did not have such an activity. The effects of tamoxifen on doxorubicin sensitivity of P388/ADR cells could not be reversed by 17 beta-estradiol. Estrogen receptors could not be demonstrated in either cell line. It is therefore suggested that the reversal of the doxorubicin-acquired resistance by the triparanol analogues is unrelated to their estrogenic or antiestrogenic activities. The possible clinical implications of these findings are discussed.
Sigma-Aldrich
Triparanol, ≥97% (HPLC), powder
Pricing and availability is not currently available.