Merck
  • Structure and properties of β-cyclodextrin/cellulose hydrogels prepared in NaOH/urea aqueous solution.
Structure and properties of β-cyclodextrin/cellulose hydrogels prepared in NaOH/urea aqueous solution.
Carbohydrate polymers (2013-04-03)
Lingzhi Zhang, Jinping Zhou, Lina Zhang
ABSTRACT
β-Cyclodextrin (β-CD)/cellulose hydrogels were prepared in NaOH/urea aqueous solution by crosslinking with epichlorohydrin. The structure and morphology of the hydrogels were characterized with Fourier transform infrared spectroscopy (FTIR), X-ray diffraction (XRD), scanning electron microscopy (SEM). The swelling test, 5-fluorouracil (5-FU) and bovine serum albumin (BSA), and aniline blue (AnB) were used to investigate the swelling capability, drug release behavior and the fluorescent property of the hydrogels. The results indicated that the swelling degree and water uptake of the hydrogels decreased with an increase of the β-CD content. The in vitro release of 5-FU and BSA of the hydrogels showed an inclusion complex formed between 5-FU and β-CD. β-CD/cellulose hydrogels adsorbed AnB lead to a fluorescence enhancement attributing to the formation of the host-guest complex between β-CD and AnB.
Sigma-Aldrich
Ascarite®, Sodium hydroxide-coated silica, 20-30 mesh
Sigma-Aldrich
Ascarite®, Sodium hydroxide-coated silica, 8-20 mesh
Supelco
Sodium hydroxide concentrate, 0.1 M NaOH in water (0.1N), Eluent concentrate for IC
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Sigma-Aldrich
Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets
Supelco
Sodium hydroxide solution, 49-51% in water, eluent for IC
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium hydroxide, reagent grade, ≥98%, pellets (anhydrous)
Sigma-Aldrich
Sodium hydroxide solution, 50% in H2O
Sigma-Aldrich
Sodium hydroxide, ACS reagent, ≥97.0%, pellets
Sigma-Aldrich
Sodium hydroxide, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, flakes
Sigma-Aldrich
Sodium hydroxide, anhydrous, free-flowing, Redi-Dri, reagent grade, ≥98%, pellets
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, powder
Sigma-Aldrich
Sodium hydroxide solution, 5.0 M
Sigma-Aldrich
Sodium hydroxide, puriss. p.a., ACS reagent, K ≤0.02%, ≥98.0% (T), pellets
Sigma-Aldrich
Sodium hydroxide solution, purum, ≥32%
Sigma-Aldrich
Sodium hydroxide, puriss., meets analytical specification of Ph. Eur., BP, NF, E524, 98-100.5%, pellets
Sigma-Aldrich
Sodium hydroxide, puriss. p.a., ACS reagent, reag. Ph. Eur., K ≤0.02%, ≥98%, pellets
Sigma-Aldrich
Sodium hydroxide, beads, 16-60 mesh, reagent grade, 97%
Sigma-Aldrich
Sodium hydroxide, pellets, semiconductor grade, 99.99% trace metals basis