Merck
  • A two-stage pretreatment approach to maximise sugar yield and enhance reactive lignin recovery from poplar wood chips.
A two-stage pretreatment approach to maximise sugar yield and enhance reactive lignin recovery from poplar wood chips.
Bioresource technology (2013-01-22)
I A Panagiotopoulos, R P Chandra, J N Saddler
ABSTRACT
A two-stage pretreatment approach, employing steam followed by organosolv treatment, was assessed for its ability to fractionate and recover most of the hemicellulose, lignin and cellulose components of poplar wood chips. A mild steaming stage was initially used to maximise hemicellulose sugar recovery, with 63% of the original xylan solubilised and recovered after this stage and close to 90% recovered in total. Rather than hindering subsequent organosolv delignification, the prior steam treatment enhanced lignin solubilisation with more than 66% of the original lignin removed after the two-stage pretreatment. The extracted lignin contained at least equal or greater amounts of functional groups as compared to the lignin solubilised after a single-stage organosolv pretreatment. More than 98% of the original cellulose was recovered after the two-stage pretreatment and 88% of the cellulose could be hydrolysed to glucose at enzyme loading of 5FPU/g cellulose after 72h.
Product Description
Supelco
Sulfuric acid, for the determination of nitrogen, ≥97.0%
Sigma-Aldrich
Sulfuric acid, 99.999%
Sigma-Aldrich
Sulfuric acid, puriss. p.a., for determination of Hg, ACS reagent, reag. ISO, reag. Ph. Eur., 95.0-97.0%
Sigma-Aldrich
Sulfuric acid solution, puriss. p.a., ≥25% (T)
Sigma-Aldrich
Sulfuric acid, puriss., meets analytical specification of Ph. Eur., BP, 95-97%
Sigma-Aldrich
Sulfuric acid, ACS reagent, 95.0-98.0%
Supelco
Sulfuric acid concentrate, 0.1 M H2SO4 in water (0.2N), eluent concentrate for IC