Merck
  • Unbalance of CB1 receptors expressed in GABAergic and glutamatergic neurons in a transgenic mouse model of Huntington's disease.
Unbalance of CB1 receptors expressed in GABAergic and glutamatergic neurons in a transgenic mouse model of Huntington's disease.
Neurobiology of disease (2011-12-31)
Valentina Chiodi, Motokazu Uchigashima, Sarah Beggiato, Antonella Ferrante, Monica Armida, Alberto Martire, Rosa Luisa Potenza, Luca Ferraro, Sergio Tanganelli, Masahiko Watanabe, Maria Rosaria Domenici, Patrizia Popoli
ABSTRACT
Cannabinoid CB1 receptors (CB1Rs) are known to be downregulated in patients and in animal models of Huntington's disease (HD). However, the functional meaning of this reduction, if any, is still unclear. Here, the effects of the cannabinoid receptor agonist WIN 55,212-2 (WIN) were investigated on striatal synaptic transmission and on glutamate and GABA release in symptomatic R6/2 mice, a genetic model of HD. The expression levels of CB1Rs in glutamatergic and GABAergic synapses were also evaluated. We found that in R6/2 mice, WIN effects on synaptic transmission and glutamate release were significantly increased with respect to wild type mice. On the contrary, a decrease in WIN-induced reduction of GABA release was found in R6/2 versus WT mice. The expression of CB1Rs in GABAergic neurons was drastically reduced, while CB1Rs levels in glutamatergic neurons were unchanged. These results demonstrate that the expression and functionality of CB1Rs are differentially affected in GABAergic and glutamatergic neurons in R6/2 mice. As a result, the balance between CB1Rs expressed by the two neuronal populations and, thus, the net effect of CB1R stimulation, is profoundly altered in HD mice.
Sigma-Aldrich
HU-210, solid (air sensitive)