Merck
  • Absorption and fluorescence spectroscopic study on complexation of Oxazine 1 Dye by calix[8]arenesulfonate.
Absorption and fluorescence spectroscopic study on complexation of Oxazine 1 Dye by calix[8]arenesulfonate.
Applied spectroscopy (2005-02-22)
Miklós Kubinyi, Tamás Vidóczy, Olívia Varga, Kornél Nagy, István Bitter
ABSTRACT
It has been established by combined absorption and fluorescence measurements that the cationic dye Oxazine 1 (OX) and the polyvalent anionic host calix[8]arenesulfonate (SCA8) form two complexes in simultaneous reactions: OX + SCA8 <--> OX.SCA8 (1), and OX.SCA8 + OX <--> OX(2).SCA8 (2). The equilibrium constants for the two reactions, as functions of the ionic strength (I), and the absorption and fluorescence spectra of the two complex species have been determined by a least-squares fitting method from the experimental data. The variations of the binding constants with the ionic strength could be described on the basis of Debye-Huckel theory. The equilibrium constants are large; their values extrapolated to I = 0 are K(1) = 5.5 x 10(6) M(-1) and K(2) = 4.4 x 10(5) M(-1). The fluorescence of OX undergoes a strong static quenching upon complexation. These results indicate that the complexes are held together by strong electrostatic forces. The addition of non-fluorescent tetramethylammonium chloride to OX-SCA8 mixtures results in a dramatic fluorescent enhancement, which demonstrates the potential applicability of this supramolecular system in fluorescence assays.
Sigma-Aldrich
Basic Blue 3, Dye content 25 %
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
250 mg
1 Estimated to ship on March 04, 2025
¥10,500