Merck
  • Photobiological properties of positively charged methylene violet analogs.
Photobiological properties of positively charged methylene violet analogs.
Photochemistry and photobiology (2000-01-29)
M A Houghtaling, R Perera, K E Owen, S Wagner, R J Kuhn, H Morrison
ABSTRACT
O-Methyl methylene violet (OMeMV), O-methyl bromomethylene violet (OMeBrMV) and O-methyl iodomethylene violet (OMeIMV) have been prepared in order to test their potential utility as anti-viral and anti-tumor phototoxic dyes. Rates of photosensitized toxicity of KB cells with 633 nm irradiation are (x 10(-19) photon-1): 2.4, 2.2, 1.9 and 0.17 for OMeIMV, OMeBrMV, methylene violet (MV) and OMeMV, respectively. Rates of photosensitized inactivation of Sindbis virus in phosphate-buffered saline with 633 nm irradiation are (x 10(-18) photon-1): 3.3, 1.8, 0.99, 0.15 for MV, OMeIMV, OMeBrMV and OMeMV, respectively. Quantum efficiencies for singlet oxygen formation were determined as OMeIMV, 0.64; OMeBrMV, 0.40; OMeMV, 0.054. Titration of the dyes with double-stranded (ds)DNA resulted in bathochromic shifts and hypochromic effects in the visible absorption spectra. Association constants for interaction of the methylated dyes with dsDNA of approximately 1 x 10(5) M-1 were determined by Scatchard analysis of equilibrium dialysis and UV absorption titration data. Photolysis of the halogenated dyes with DNA under argon led to covalent bond formation with the nucleic acid; there was no evidence of covalent binding in the dark.
Sigma-Aldrich
Methylene Violet (Bernthsen), certified by the Biological Stain Commission, Dye content ≥65 %