Merck
  • Fast product formation and slow product release are important features in a hysteretic reaction mechanism of glutathione transferase T2-2.
Fast product formation and slow product release are important features in a hysteretic reaction mechanism of glutathione transferase T2-2.
Biochemistry (1999-08-06)
P Jemth, B Mannervik
ABSTRACT
The reaction mechanism of rat glutathione transferase T2-2 has been studied using pre-steady-state and steady-state kinetics. Several parts of the catalytic cycle including binding of substrates, product formation, and product release were investigated. Under saturating conditions, a two-step product release was found to be rate limiting in the enzyme-catalyzed reactions between the nucleophilic substrate glutathione and either of the two electrophilic substrates 1-menaphthyl sulfate and 4-nitrobenzyl chloride. The rate constant for pre-steady-state product formation on rat glutathione transferase T2-2 has an observed pK(a) value of 5.7 apparently due to ionization of the sulfhydryl group of glutathione. This rate constant is approximately 2 orders of magnitude higher than k(cat) at pH values of >6. It can be predicted from the pH dependence that product formation would be the sole rate-limiting step at pH values of <3. A hysteretic mechanism of rGST T2-2 is proposed based on a slow conformational transition detected in pre-steady-state displacement experiments.
Sigma-Aldrich
4-Nitrobenzyl chloride, 99%