Merck
  • Cognitive-enhancing and antioxidant activities of iridoid glycosides from Scrophularia buergeriana in scopolamine-treated mice.
Cognitive-enhancing and antioxidant activities of iridoid glycosides from Scrophularia buergeriana in scopolamine-treated mice.
European journal of pharmacology (2008-05-09)
Eun Ju Jeong, Ki Yong Lee, Seung Hyun Kim, Sang Hyun Sung, Young Choong Kim
ABSTRACT
The cognitive-enhancing activities of E-harpagoside and 8-O-E-p-methoxycinnamoylharpagide (MCA-Hg) isolated from Scrophularia buergeriana were evaluated in scopolamine-induced amnesic mice by the Morris water maze and by passive avoidance tests. E-harpagoside and MCA-Hg significantly improved the impairment of reference memory induced by scopolamine in the Morris water maze test. The mean escape latency, the mean path length and swimming movement were also improved by both compounds. In passive avoidance test, E-harpagoside and MCA-Hg (2 mg/kg body weight, p.o.) significantly ameliorated scopolamine-induced amnesia by as much as 70% of the level found in normal control mice. Donepezil, an acetylcholinesterase inhibitor and the most widely used drug for AD treatment was employed as a positive control. The activity of acetylcholinesterase was inhibited significantly by E-harpagoside or MCA-Hg within the cortex and hippocampus to a level similar to that observed in mice treated with donepezil (2 mg/kg body weight, p.o.). Moreover, treatment with E-harpagoside or MCA-Hg to scopolamine-induced amnesic mice significantly decreased TBARS level which was accompanied by an increase in the activities or contents of glutathione reductase, SOD and reduced GSH. We believe these data demonstrate that E-harpagoside or MCA-Hg exerted potent cognitive-enhancing activity through both anti-acetylcholinesterase and antioxidant mechanisms.
Product Description
Harpagoside, primary reference standard
SKU
Pack Size
Availability
Price
Quantity
Supelco
Harpagoside, analytical standard