Merck
  • Optimization of solid-phase extraction for the liquid chromatography-mass spectrometry analysis of harpagoside, 8-para-coumaroyl harpagide, and harpagide in equine plasma and urine.
Optimization of solid-phase extraction for the liquid chromatography-mass spectrometry analysis of harpagoside, 8-para-coumaroyl harpagide, and harpagide in equine plasma and urine.
Journal of chromatographic science (2008-03-28)
Cyril Colas, Patrice Garcia, Marie-Agnès Popot, Yves Bonnaire, Stéphane Bouchonnet
ABSTRACT
Solid-phase extraction cartridges among those usually used for screening in horse doping analyses are tested to optimize the extraction of harpagoside (HS), harpagide (HG), and 8-para-coumaroyl harpagide (8PCHG) from plasma and urine. Extracts are analyzed by liquid chromatography coupled with multi-step tandem mass spectrometry. The extraction process retained for plasma applies BondElut PPL cartridges and provides extraction recoveries between 91% and 93%, with RSD values between 8 and 13% at 0.5 ng/mL. Two different procedures are needed to extract analytes from urine. HS and 8PCHG are extracted using AbsElut Nexus cartridges, with recoveries of 85% and 77%, respectively (RSD between 7% and 19%). The extraction of HG involves the use of two cartridges: BondElut PPL and BondElut C18 HF, with recovery of 75% and RSD between 14% and 19%. The applicability of the extraction methods is determined on authentic equine plasma and urine samples after harpagophytum or harpagoside administration.
Product Description
Harpagoside, primary reference standard
Pricing and availability is not currently available.
Supelco
Harpagoside, analytical standard
Pricing and availability is not currently available.