Merck
  • Gas-liquid chromatographic properties of positional isomers of methyl thia, selena, and tellura laurate analogs.
Gas-liquid chromatographic properties of positional isomers of methyl thia, selena, and tellura laurate analogs.
Lipids (1997-06-01)
M S Jie, O Bakare, Y K Cheung, S H Chau
ABSTRACT
Gas-liquid chromatographic analyses of three complete series of synthetic positional isomers of methyl thia, selena, and tellura laurate analogs were carried on a nonpolar (SE-30) and a polar (SP-2330) stationary phase. The average ECL (equivalent chain length) values of the thia, selena, and tellura laurate on SE-30 stationary phase were 13.8, 14.8, and 15.7, respectively, while on SP-2330 the average values for the same series were 17.1, 19.0, and 19.1, respectively. Positional isomers with the heteroatom at the 2-position exhibited the lowest ECL values, while those with the heteroatom at the omega-1 position gave the highest ECL values and were readily separated from the other positional isomers of the same series of analogs by this technique.
Product Description
Supelco
SP®-2330 Capillary GC Column, L × I.D. 30 m × 0.25 mm, df 0.20 μm
Supelco
SP®-2330 Capillary GC Column, L × I.D. 30 m × 0.32 mm, df 0.20 μm
Supelco
SP®-2330 Capillary GC Column, L × I.D. 60 m × 0.32 mm, df 0.20 μm
Supelco
SP®-2330 Capillary GC Column, L × I.D. 15 m × 0.25 mm, df 0.20 μm
Supelco
SP®-2330 Capillary GC Column, L × I.D. 60 m × 0.25 mm, df 0.20 μm
Supelco
Methyl laurate, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Methyl laurate, 99.5%
Sigma-Aldrich
Methyl laurate, ≥98%, FG
Supelco
Methyl dodecanoate, analytical standard