Merck
  • Comparison of IR- and UV-matrix-assisted laser desorption/ionization mass spectrometry of oligodeoxynucleotides.
Comparison of IR- and UV-matrix-assisted laser desorption/ionization mass spectrometry of oligodeoxynucleotides.
Nucleic acids research (1994-07-11)
E Nordhoff, F Kirpekar, M Karas, R Cramer, S Hahner, F Hillenkamp, K Kristiansen, P Roepstroff, A Lezius
ABSTRACT
UV-matrix assisted laser desorption/ionization mass spectrometry (UV-MALDI-MS) with 3-hydroxypicolinic acid as matrix and IR-MALDI-MS with succinic acid as matrix have proved their feasibility for highly accurate and sensitive mass determination of nucleic acids (DNA and RNA). In this work, a detailed comparison of these two MALDI-methods and between positive- and negative ion mass spectra for the analysis of oligodeoxynucleotides is undertaken. Mass spectra of DNA sequences with up to 40 nucleotides are shown. Both linear and reflectron time-of-flight mass analyzers were used within this study and are compared for their potential in the MALDI analysis of oligodeoxynucleotides. The role of molecule-ion fragmentation is also discussed.
Product Description
Supelco
3-Hydroxypicolinic acid, matrix substance for MALDI-MS, ≥99.0% (HPLC)
Sigma-Aldrich
3-Hydroxypyridine-2-carboxylic acid, 98%