Merck
Allergy to phenyl salicylate.
Contact dermatitis (1981-07-01)
C D Calnan, E Cronin, R J Rycroft
ABSTRACT
Six cases of contact dermatitis from a lip salve are described. Five were allergic to phenyl salicylate and one to geraniol in the fragrance. The dermatitis spread in a ring around the mouth. Phenyl salicylate has been removed from the formulation.
Sigma-Aldrich
Phenyl salicylate, ReagentPlus®, 99%
Supelco
Phenyl salicylate melting point standard, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Phenyl salicylate, ≥99%, FG
Supelco
Mettler-Toledo Calibration substance ME 30034252, Phenyl salicylate, traceable to primary standards