Merck
  • Preparation of 8,5'-O-cycloadenosine derivatives by reaction of 2',3'-O-isopropylideneadenosine with phenyl chloroformate or p-toluyl chloride.
Preparation of 8,5'-O-cycloadenosine derivatives by reaction of 2',3'-O-isopropylideneadenosine with phenyl chloroformate or p-toluyl chloride.
Nucleic acids symposium series (1984-01-01)
K Anzai, J Uzawa
ABSTRACT
Reaction of 2',3'-O-isopropylideneadenosine with p-toluyl chloride in a mixture of methylene chloride and triethylamine afforded 2',3'-O-isopropylidene-N,N,5'-O-tri-p-toluyladenosine (8)(30%), 8,5'-O-cycloadenosine derivatives 9 (34%) and 10 (11%), and a cyanoimidazole nucleoside 11 (12%).
Sigma-Aldrich
2′,3′-O-Isopropylideneadenosine, 98%
Sigma-Aldrich
Phenyl chloroformate, 99%
Sigma-Aldrich
Phenyl chloroformate, 97%