Merck
  • Metabolism of 5-methyl-2-furaldehyde in rat. III. Identification and determination of 5-methyl-2-furylmethylketone.
Metabolism of 5-methyl-2-furaldehyde in rat. III. Identification and determination of 5-methyl-2-furylmethylketone.
Xenobiotica; the fate of foreign compounds in biological systems (1988-08-01)
J Jodynis-Liebert
ABSTRACT
1. Continuous extraction, column chromatography and t.l.c. were employed to isolate a minor metabolite of 5-methyl-2-furaldehyde from rat urine. 2. The metabolite was identified by mass spectrometry and independent synthesis as 5-methyl-2-furylmethylketone. 3. A method for quantitative determination of the metabolite in urine was devised. About 7% of the parent compound was metabolized to 5-methyl-2-furylmethylketone.
Sigma-Aldrich
2-Acetyl-5-methylfuran, 98%
Sigma-Aldrich
5-Methylfurfural, ReagentPlus®, 99%
Sigma-Aldrich
2-Acetyl-5-methylfuran, ≥98%, FG
Sigma-Aldrich
5-Methylfurfural, ≥98%, FG