Merck
  • [Prohepsidin in diagnostic of iron deficiency in patients with chronic cardiac failure and anemia].
[Prohepsidin in diagnostic of iron deficiency in patients with chronic cardiac failure and anemia].
Klinicheskaia laboratornaia diagnostika (2013-07-03)
N M Makarova, I I Shaposhnik
ABSTRACT
The sample for examination included 52 patients with chronic cardiac failure and 26 patients with anemia amongst. The expected increase of production of prohepsidin against the background of increased serum concentrations of anti-inflammatory cytokines was not detected. The tendency to decrease level of prohepsidin alongside with decrease of content of serum iron and ferritin in blood serum testified depletion of iron resources in organism.
Sigma-Aldrich
Iron, puriss. p.a., carbonyl-Iron powder, low in magnesium and manganese compounds, ≥99.5% (RT)
Iron, IRMM®, certified reference material, 0.5 mm wire
Sigma-Aldrich
Iron, foil, thickness 0.1 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Iron, powder, −325 mesh, 97%
Sigma-Aldrich
Iron, chips, 99.98% trace metals basis
Sigma-Aldrich
Ferritin from human spleen, Type V, 10 μg/mL in 0.15 M NaCl, 10 mM Tris, pH 8.0, containing 0.1% sodium azide
Sigma-Aldrich
Ferritin from equine spleen, Type I, saline solution
Sigma-Aldrich
Iron, granular, 10-40 mesh, >99.99% trace metals basis
Sigma-Aldrich
Iron, wire, diam. 1.0 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Iron, ≥99%, reduced, powder (fine)
Sigma-Aldrich
Iron, foil, thickness 0.25 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Ferritin from human liver, Type IV, 10 μg/mL
Sigma-Aldrich
Carbonyl iron, ≥97% Fe basis