Merck
  • Functionalization of mesoporous silica nanoparticles for targeting, biocompatibility, combined cancer therapies and theragnosis.
Functionalization of mesoporous silica nanoparticles for targeting, biocompatibility, combined cancer therapies and theragnosis.
Journal of nanoscience and nanotechnology (2013-06-15)
Yaswanth Kuthati, Ping-Jyun Sung, Ching-Feng Weng, Chung-Yuan Mou, Chia-Hung Lee
ABSTRACT
The advent of Nanotechnology has paved a way for improved disease treatment strategies, the most noteworthy being the mesoporous silica nanoparticles (MSNs) which have gained much recent attention in the field of cancer therapy and its diagnosis. The flaws of the current-day strategies can be overcome by this superior technology through its targeting ability in delivering drugs and image able agents specifically to the tumor sites. MSNs have unique biocompatibility features, its high surface area which contributes in large amount of drug loading and its facility to monitor size and shape of the nanoparticles are few of the positives which makes this technology an enormous asset for the field of Nanotechnology. This review paper is structured in such a way wherein we initially have discussed about the synthesis methods and various functionalization approaches for MSN followed by the different methods used for targeting cancer cells and the latest advances in controlled drug release. Some of the highlights of this review are the biocompatibility of MSNs, in vivo results of MSNs on cancer therapy. This review paper also shortly discuss about combined cancer therapies to overcome the challenges in current-day cancer treatment. Finally, we converge briefly on the recent advancements in the use of hybrid MSNs for obtaining multiple functions.
Sigma-Aldrich
LUDOX® HS-40 colloidal silica, 40 wt. % suspension in H2O
Pricing and availability is not currently available.
Sigma-Aldrich
LUDOX® AM colloidal silica, 30 wt. % suspension in H2O
Pricing and availability is not currently available.
Sigma-Aldrich
LUDOX® CL-X colloidal silica, 45 wt. % suspension in H2O
Pricing and availability is not currently available.
Sigma-Aldrich
LUDOX® AS-40 colloidal silica, 40 wt. % suspension in H2O
Pricing and availability is not currently available.
Sigma-Aldrich
LUDOX® TMA colloidal silica, 34 wt. % suspension in H2O
Pricing and availability is not currently available.
Sigma-Aldrich
LUDOX® AS-30 colloidal silica, 30 wt. % suspension in H2O
Pricing and availability is not currently available.
Sigma-Aldrich
LUDOX® TM-50 colloidal silica, 50 wt. % suspension in H2O
Pricing and availability is not currently available.
Sigma-Aldrich
Silica, nanopowder, 99.8% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Silica
Pricing and availability is not currently available.
Sigma-Aldrich
LUDOX® SM colloidal silica, 30 wt. % suspension in H2O
Pricing and availability is not currently available.
Sigma-Aldrich
LUDOX® HS-30 colloidal silica, 30 wt. % suspension in H2O
Pricing and availability is not currently available.
Sigma-Aldrich
LUDOX® TM-40 colloidal silica, 40 wt. % suspension in H2O
Pricing and availability is not currently available.
Sigma-Aldrich
Silicon dioxide, alumina doped, nanoparticles, dispersion, <50 nm particle size, 20 wt. % in H2O, ≥99.9% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Silica, mesostructured, MSU-F (cellular foam)
Pricing and availability is not currently available.
Sigma-Aldrich
LUDOX® CL colloidal silica, 30 wt. % suspension in H2O
Pricing and availability is not currently available.
Sigma-Aldrich
LUDOX® LS colloidal silica, 30 wt. % suspension in H2O
Pricing and availability is not currently available.
Sigma-Aldrich
Silicon dioxide, nanopowder (spherical, porous), 5-20 nm particle size (TEM), 99.5% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Silica, mesostructured, MCM-41 type (hexagonal)
Pricing and availability is not currently available.
Sigma-Aldrich
Silica, nanoparticles, mesoporous, 200 nm particle size, pore size 4 nm
Pricing and availability is not currently available.
Sigma-Aldrich
Silicon dioxide, nanopowder, 10-20 nm particle size (BET), 99.5% trace metals basis
Pricing and availability is not currently available.