Merck
Defining the RGG/RG motif.
Molecular cell (2013-06-12)
Palaniraja Thandapani, Timothy R O'Connor, Timothy L Bailey, Stéphane Richard
ABSTRACT
Motifs rich in arginines and glycines were recognized several decades ago to play functional roles and were termed glycine-arginine-rich (GAR) domains and/or RGG boxes. We review here the evolving functions of the RGG box along with several sequence variations that we collectively term the RGG/RG motif. Greater than 1,000 human proteins harbor the RGG/RG motif, and these proteins influence numerous physiological processes such as transcription, pre-mRNA splicing, DNA damage signaling, mRNA translation, and the regulation of apoptosis. In particular, we discuss the role of the RGG/RG motif in mediating nucleic acid and protein interactions, a function that is often regulated by arginine methylation and partner-binding proteins. The physiological relevance of the RGG/RG motif is highlighted by its association with several diseases including neurological and neuromuscular diseases and cancer. Herein, we discuss the evidence for the emerging diverse functionality of this important motif.
Product Description
Supelco
L-Arginine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Arginine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Arginine, 99%, FCC, FG
SAFC
L-Arginine
Sigma-Aldrich
L-Arginine, reagent grade, ≥98%
Sigma-Aldrich
L-Arginine, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 98.5-101.0%