Merck
  • A facile route towards the synthesis of polystyrene/zinc oxide nanocomposites.
A facile route towards the synthesis of polystyrene/zinc oxide nanocomposites.
Journal of nanoscience and nanotechnology (2013-05-08)
Long Giang Bach, M Rafiqul Islam, Yong Hun Kim, Sung Deok Seo, Chan Park, Hyun Gyu Kim, Kwon Taek Lim
ABSTRACT
ZnO nanoparticles were covalently wrapped by polystyrene (PS) through surface thiol-lactam initiated radical polymerization using the grafting from approach. The surface of ZnO nanoparticles was initially modified by 3-mercapto propyltrimethoxysilane to afford thiol functionalized ZnO nanoparticles (ZnO-SH). The controlled radical polymerization of styrene (St) was subsequently accomplished by using an initiating system of ZnO-SH and butyrolactam. FT-IR, XPS, XRD, TGA, DSC, TEM and DLS were employed to investigate the chemical structure, morphology, thermal properties, the size and size distribution of nanocomposites. The dispersibility of ZnO nanoparticles was observed to be significantly improved upon functionalization by PS brushes. The controlled nature of the surface initiated thiol-lactam aided polymerization of St from the ZnO nanoparticles surface was confirmed by GPC analysis.
Sigma-Aldrich
Zinc oxide, ReagentPlus®, powder, <5 μm particle size, 99.9%
Sigma-Aldrich
Zinc oxide, puriss., meets analytical specification of Ph. Eur., BP, USP, 99-100.5% (calc. for dried substance)
Sigma-Aldrich
Zinc oxide, puriss. p.a., ACS reagent, ≥99.0% (KT)
Supelco
Zinc oxide, analytical standard
Sigma-Aldrich
Zinc oxide, 99.99% trace metals basis
Sigma-Aldrich
Zinc oxide, dispersion, nanoparticles, <100 nm particle size (TEM), ≤40 nm avg. part. size (APS), 20 wt. % in H2O
Sigma-Aldrich
Zinc oxide, nanopowder, <50 nm particle size (BET), >97%
Sigma-Aldrich
Zinc oxide, nanopowder, <100 nm particle size
Sigma-Aldrich
Zinc oxide, 99.999% trace metals basis