Merck
  • Overcoming drug-resistant herpes simplex virus (HSV) infection by a humanized antibody.
Overcoming drug-resistant herpes simplex virus (HSV) infection by a humanized antibody.
Proceedings of the National Academy of Sciences of the United States of America (2013-04-10)
Adalbert Krawczyk, Michaela A E Arndt, Ludger Grosse-Hovest, Wilko Weichert, Bernd Giebel, Ulf Dittmer, Hartmut Hengel, Dirk Jäger, Karl E Schneweis, Anna M Eis-Hübinger, Michael Roggendorf, Jürgen Krauss
ABSTRACT
Despite the availability of antiviral chemotherapy, herpes simplex virus type 1 (HSV-1) and type 2 (HSV-2) infections remain a severe global health problem. Of particular concern is the growing incidence of drug resistance in immunocompromised patients, which stresses the urgency to develop new effective treatment alternatives. We have developed a humanized monoclonal antibody (mAb hu2c) that completely abrogates viral cell-to-cell spread, a key mechanism by which HSV-1/2 escapes humoral immune surveillance. Moreover, mAb hu2c neutralized HSV fully independent of complement and/or immune effector cell recruitment in a highly efficient manner. Prophylactic and therapeutic administration of mAb hu2c completely prevented infection-related mortality of severely immunodeficient mice being challenged with a lethal dose of HSV-1. The high neutralization capacity of mAb hu2c was fully maintained toward clinical HSV isolates being multiresistant to standard antiviral drugs, and infection was fully resolved in 7/8 nonobese diabetic/SCID mice being infected with a multidrug resistant HSV-1 patient isolate. Immunohistochemical studies revealed no significant cross-reactivity of the antibody toward human tissues. These features warrant further clinical development of mAb hu2c as an immunotherapeutic compound for the management of severe and particularly drug-resistant HSV infections.
Product Description
Aciclovir, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Acycloguanosine, ≥99% (HPLC), powder
Supelco
Acyclovir, Pharmaceutical Secondary Standard; Certified Reference Material