- The molybdenum cofactor.
ABSTRACT
The transition element molybdenum needs to be complexed by a special cofactor to gain catalytic activity. Molybdenum is bound to a unique pterin, thus forming the molybdenum cofactor (Moco), which, in different variants, is the active compound at the catalytic site of all molybdenum-containing enzymes in nature, except bacterial molybdenum nitrogenase. The biosynthesis of Moco involves the complex interaction of six proteins and is a process of four steps, which also require iron, ATP, and copper. After its synthesis, Moco is distributed, involving Moco-binding proteins. A deficiency in the biosynthesis of Moco has lethal consequences for the respective organisms.
Product Description
Molybdenum, foil, not light tested, 50x50mm, thickness 0.0125mm, 99.9%
Molybdenum, rod, 1000mm, diameter 2.0mm, centerless ground, 99.9%
Molybdenum, foil, light tested, 25x25mm, thickness 0.005mm, 99.9%
Molybdenum, wire reel, 100m, diameter 0.2mm, as drawn, 99.95%
Molybdenum, foil, light tested, 25x25mm, thickness 0.004mm, 99.9%
Molybdenum, microfoil, disks, 25mm, thinness 1.0μm, specific density 1022μg/cm2, permanent mylar 3.5μm support, 99.9%
Molybdenum, foil, not light tested, 25x25mm, thickness 0.006mm, 99.9%
Molybdenum, foil, light tested, 50x50mm, thickness 0.006mm, 99.9%
Molybdenum, wire reel, 1m, diameter 0.013mm, hard, 99.8+%
Molybdenum, foil, 100x100mm, thickness 0.75mm, annealed, 99.9%
Molybdenum, wire reel, 10m, diameter 0.01mm, hard, 99.8+%
Molybdenum, wire reel, 100m, diameter 0.025mm, hard, 99.95%
Molybdenum, wire reel, 100m, diameter 0.013mm, hard, 99.8+%
Molybdenum, wire reel, 10m, diameter 0.30mm, annealed, 99.95%
Molybdenum, wire reel, 100m, diameter 1.25mm, annealed, 99.95%
Molybdenum, foil, 100x100mm, thickness 0.15mm, annealed, 99.9%
Molybdenum, foil, 6mm disks, thickness 0.0125mm, 99.9%
Molybdenum, foil, 4mm disks, thickness 0.005mm, 99.9%
Molybdenum, foil, 6mm disks, thickness 0.015mm, annealed, 99.9%
Molybdenum, foil, 4mm disks, thickness 0.30mm, annealed, 99.9%
Molybdenum, foil, 50mm disks, thickness 0.025mm, annealed, 99.9%
Molybdenum, foil, 15mm disks, thickness 0.025mm, annealed, 99.9%
Molybdenum, foil, light tested, 25x25mm, thickness 0.05mm, annealed, 99.9%