Merck
Treatment course with antidepressant therapy in late-life depression.
The American journal of psychiatry (2013-03-28)
Yvette I Sheline, Brianne M Disabato, Jennifer Hranilovich, Carrie Morris, Gina D'Angelo, Carl Pieper, Tommaso Toffanin, Warren D Taylor, James R MacFall, Consuelo Wilkins, Deanna M Barch, Kathleen A Welsh-Bohmer, David C Steffens, Ranga R Krishnan, P Murali Doraiswamy
ABSTRACT
In order to assess the effect of gray matter volumes and cortical thickness on antidepressant treatment response in late-life depression, the authors examined the relationship between brain regions identified a priori and Montgomery-Åsberg Depression Rating Scale (MADRS) scores over the course of an antidepressant treatment trial. In a nonrandomized prospective trial, 168 patients who were at least 60 years of age and met DSM-IV criteria for major depression underwent MRI and were enrolled in a 12-week treatment study. Exclusion criteria included cognitive impairment or severe medical disorders. The volumes or cortical thicknesses of regions of interest that differed between the depressed group and a comparison group (N=50) were determined. These regions of interest were used in analyses of the depressed group to predict antidepressant treatment outcome. Mixed-model analyses adjusting for age, education, age at depression onset, race, baseline MADRS score, scanner, and interaction with time examined predictors of MADRS scores over time. Smaller hippocampal volumes predicted a slower response to treatment. With the inclusion of white matter hyper-intensity severity and neuropsychological factor scores, the best model included hippocampal volume and cognitive processing speed to predict rate of response over time. A secondary analysis showed that hippocampal volume and frontal pole thickness differed between patients who achieved remission and those who did not. These data expand our understanding of the prediction of treatment course in late-life depression. The authors propose that the primary variables of hippocampal volume and cognitive processing speed, subsuming other contributing variables (episodic memory, executive function, language processing) predict antidepressant response.
Product Description
Sertraline for system suitability, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.
Sertraline for peak identification, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.
Sigma-Aldrich
Sertraline hydrochloride, ≥98% (HPLC)
Pricing and availability is not currently available.
Supelco
Sertraline hydrochloride solution, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®
Pricing and availability is not currently available.