Merck
  • Effect of intrathecal magnesium in the presence or absence of local anaesthetic with and without lipophilic opioids: a systematic review and meta-analysis.
Effect of intrathecal magnesium in the presence or absence of local anaesthetic with and without lipophilic opioids: a systematic review and meta-analysis.
British journal of anaesthesia (2013-03-28)
A P Morrison, J M Hunter, S H Halpern, A Banerjee
ABSTRACT
Spinal anaesthesia is the primary anaesthetic technique for many types of surgery. Adjuncts to the local anaesthetics (LA) used in spinal anaesthesia can exhibit undesirable side-effects, limiting their use, but magnesium may have advantages in this respect. We sought randomized control trials (RCTs) in patients undergoing all types of surgery and in women in labour to compare the effect of intrathecal magnesium sulphate ± LA ± lipophilic opioid (experimental group) with the use of either intrathecal lipophilic opioids ± LA or LA only (control group). The primary outcome was the duration of spinal anaesthesia. Secondary outcomes were: onset and time to maximal sensory blockade, onset of motor block, and duration of sensory and motor blockade. We found 15 RCTs comprising 980 patients. The duration of spinal anaesthesia was significantly increased in the experimental group [standardized mean difference (SMD) -1.05 (-1.70, -0.41) (P = 0.001)], compared with the control group. This increased duration of spinal anaesthesia was seen in non-obstetric studies, SMD -1.38 (-2.11, -0.66) (P = 0.0002), but not in obstetric studies, SMD -0.55 (-1.87, 0.77) (P = 0.41). There was no delay in the onset of sensory or motor blockade. The incidence of hypotension and pruritus was similar in both groups. Heterogeneity was high in all outcome measures. The duration of spinal anaesthesia may be increased by the addition of magnesium to lipophilic opioids ± LA.
Sigma-Aldrich
Magnesium sulfate heptahydrate, BioUltra, ≥99.5% (KT)
Sigma-Aldrich
Magnesium sulfate heptahydrate, BioReagent, for molecular biology, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Magnesium sulfate heptahydrate, BioXtra, ≥99.0%
Sigma-Aldrich
Magnesium sulfate heptahydrate, puriss. p.a., ACS reagent, ≥99.0% (KT)
Sigma-Aldrich
Magnesium sulfate heptahydrate, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Magnesium sulfate heptahydrate, 99.5-100.5% (calc. to the dried substance), meets analytical specification of Ph. Eur., BP,USP, FCC
Sigma-Aldrich
Magnesium sulfate heptahydrate, ACS reagent, ≥98%
Sigma-Aldrich
Magnesium sulfate, anhydrous, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Magnesium sulfate, puriss. p.a., drying agent, anhydrous, ≥98.0% (KT), powder (very fine)
Sigma-Aldrich
Magnesium sulfate, anhydrous, reagent grade, ≥97%
Sigma-Aldrich
Magnesium sulfate solution, BioUltra, for molecular biology
Sigma-Aldrich
Magnesium sulfate, ≥99.99% trace metals basis
Sigma-Aldrich
Magnesium sulfate, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Magnesium sulfate solution, for molecular biology, 1.00 M±0.04 M