Merck
  • Survival and transfer of microorganisms from kitchen sponges to surfaces of stainless steel and polyethylene.
Survival and transfer of microorganisms from kitchen sponges to surfaces of stainless steel and polyethylene.
Journal of infection in developing countries (2013-03-16)
Eliandra Mirlei Rossi, Diane Scapin, Eduardo César Tondo
ABSTRACT
Contaminated sponges might lead to cross-contamination in kitchens since they can transfer microorganisms to surfaces where microorganisms can survive for hours or days and contaminate food. The main objective of this study was to evaluate the transfer and the survival of bacteria from kitchen sponges to surfaces of AISI 316 stainless steel and polyethylene. Twenty-four sponges were collected from industrial kitchens in the state of Rio Grande do Sul and aseptically split into two equal parts. One part was subjected to enumeration of heterotrophic microorganisms, faecal coliforms, coagulase-positive Staphylococcus and search detection of Salmonella enterica. The other part was rubbed on surfaces of AISI 316 stainless steel (12 sponges) or polyethylene (12 sponges). The transfer and survival of microorganisms was quantified by swab collection and pour-plate method using plate count agar. All sponges were contaminated by heterotrophic microorganisms (average of 6.8 log CFU/sponge) and 83.3% with faecal coliforms (average of 5 log CFU/sponge). None of the sponges were contaminated by S. enterica and/or coagulase-positive Staphylococcus. The average transfer of microorganisms varied between 3.3 and 5.5 log CFU/cm2 for stainless steel and from 3.5 to 5.6 log CFU/cm2 for polyethylene. Although the survival rate decreased over time, more than 1 log CFU/cm2 of heterotrophic microorganisms survived after 24 hours on both surfaces. The sponges used in food services were significantly contaminated and could transfer large amounts of microorganisms to surfaces of AISI 316 stainless steel and polyethylene.
Sigma-Aldrich
Polyethylene, average Mw ~4,000 by GPC, average Mn ~1,700 by GPC
Sigma-Aldrich
Polyethylene, Medium density
Sigma-Aldrich
Polyethylene, Ultra-high molecular weight, average Mw 3,000,000-6,000,000
Sigma-Aldrich
Polyethylene, Ultra-high molecular weight, surface-modified, powder, 125 μm avg. part. size
Sigma-Aldrich
Polyethylene, Ultra-high molecular weight, surface-modified, powder, 34-50 μm particle size
Sigma-Aldrich
Polyethylene, low density, melt index 25 g/10 min (190°C/2.16kg)
Sigma-Aldrich
Polyethylene, Linear low density, melt index 1.0 g/10 min (190°C/2.16kg)
Sigma-Aldrich
Polyethylene, High density, melt index 12 g/10 min (190 °C/2.16kg)
Sigma-Aldrich
Polyethylene, High density, melt index 2.2 g/10 min (190 °C/2.16kg)
Supelco
Polyethylene, analytical standard, for GPC, 2,000
Polyethylene (LDPE), ERM®, certified reference material