Merck
  • A consensus envelope protein domain III can induce neutralizing antibody responses against serotype 2 of dengue virus in non-human primates.
A consensus envelope protein domain III can induce neutralizing antibody responses against serotype 2 of dengue virus in non-human primates.
Archives of virology (2013-03-05)
Hsin-Wei Chen, Shih-Jen Liu, Yi-Shiuan Li, Hsueh-Hung Liu, Jy-Ping Tsai, Chen-Yi Chiang, Mei-Yu Chen, Chyi-Sing Hwang, Chin-Cheng Huang, Hui-Mei Hu, Han-Hsuan Chung, Sze-Hsien Wu, Pele Chong, Chih-Hsiang Leng, Chien-Hsiung Pan
ABSTRACT
We have previously demonstrated that vaccination with a subunit dengue vaccine containing a consensus envelope domain III with aluminum phosphate elicits neutralizing antibodies against all four serotypes of dengue virus in mice. In this study, we evaluated the immunogenicity of the subunit dengue vaccine in non-human primates. After vaccination, monkeys that received the subunit vaccine with aluminum phosphate developed a significantly strong and long-lasting antibody response. A specific T cell response with cytokine production was also induced, and this correlated with the antibody response. Additionally, neutralizing antibodies against serotype 2 were detected in two of three monkeys. The increase in serotype-2-specific antibody titers and avidity observed in these two monkeys suggested that a serotype-2-biased antibody response occurs. These data provide evidence that a protective neutralizing antibody response was successfully elicited in non-human primates by the dengue subunit vaccine with aluminum phosphate adjuvant.
Sigma-Aldrich
Aluminum phosphate monobasic solution, ~50% in H2O
Sigma-Aldrich
Aluminum phosphate monobasic, purum, ≥95.0% (gravimetric)
Sigma-Aldrich
Aluminum phosphate, 99.99% trace metals basis
Sigma-Aldrich
Aluminum phosphate, reagent grade