Merck
  • The onset of Triton X-100 solubilization of sphingomyelin/ceramide bilayers: effects of temperature and composition.
The onset of Triton X-100 solubilization of sphingomyelin/ceramide bilayers: effects of temperature and composition.
Chemistry and physics of lipids (2013-03-05)
Hasna Ahyayauch, Cristina Arnulphi, Jesús Sot, Alicia Alonso, Félix M Goñi
ABSTRACT
The early stages of Triton X-100 solubilization of bilayers consisting of sphingomyelin/ceramide (SM/Cer) mixtures have been studied using a combination of calorimetric and spectroscopic techniques. Compositions based on sphingomyelin, containing up to 30 mol% Cer, at 4, 20 and 50°C have been examined. The presence of Cer does not modify the affinity (in terms of ΔG of binding per mol total lipid) of the SM-based bilayers for Triton X-100, although it does increase the amount of detergent required for the onset of solubilization. At 50°C more detergent was required to solubilize the SM/Cer bilayers than at 20°C. The data can be rationalized in terms of lipid and detergent geometries and interactions (Lichtenberg et al., 2013).
Sigma-Aldrich
Triton X-100 solution, BioUltra, for molecular biology, ~10% in H2O
Sigma-Aldrich
Triton X-305 solution, 70% in H2O
Sigma-Aldrich
Triton X-102
Octoxinol 10, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Triton X-45
Sigma-Aldrich
Triton X-100, peroxide- and carbonyl-free
Sigma-Aldrich
Triton X-100, BioXtra
Sigma-Aldrich
Triton X-100, for molecular biology