Merck
  • Macrolides inhibit Fusobacterium nucleatum-induced MUC5AC production in human airway epithelial cells.
Macrolides inhibit Fusobacterium nucleatum-induced MUC5AC production in human airway epithelial cells.
Antimicrobial agents and chemotherapy (2013-02-06)
Kentaro Nagaoka, Katsunori Yanagihara, Yosuke Harada, Koichi Yamada, Yohei Migiyama, Yoshitomo Morinaga, Hiroo Hasegawa, Koichi Izumikawa, Hiroshi Kakeya, Masaharu Nishimura, Shigeru Kohno
ABSTRACT
Fusobacterium nucleatum is one of the most common anaerobic bacteria in periodontitis and is responsible for several extraoral infections, including respiratory tract diseases. In this study, we examined whether F. nucleatum induces mucin secretion in airway epithelial cells. We also examined the effects of macrolides on F. nucleatum-induced mucus production compared with the effects of other antibiotics that exert anti-anaerobic activities. The production of MUC5AC, the major core protein of mucin secreted from the airway surface epithelium, in bronchial epithelial cells after stimulation with culture supernatants (Sup) of F. nucleatum was analyzed by performing enzyme-linked immunosorbent assay and quantitative RT-PCR. The cell-signaling pathway of F. nucleatum Sup stimulation was also analyzed by Western blotting. For inhibition studies, cells were treated with azithromycin, clarithromycin, clindamycin (CLDM), and metronidazole (MTZ). The F. nucleatum Sup induced NCI-H292 cells to express MUC5AC at both the protein level and the mRNA level in both a time- and dose-dependent manner. Macrolides inhibited F. nucleatum Sup-induced MUC5AC production, while CLDM and MTZ were less effective. F. nucleatum Sup induced the phosphorylation of extracellular signal-regulated kinase 1/2 (ERK1/2), and this induction was suppressed by macrolides. F. nucleatum Sup-induced MUC5AC production was blocked by the ERK pathway inhibitor U0126. F. nucleatum is likely to contribute to excessive mucin production, which suggests that periodontitis may correlate with the pathogenesis of chronic respiratory tract infection. Macrolides seem to reduce this mucin production and might represent an additional means of therapeutic intervention for F. nucleatum respiratory tract infections other than CLDM and MTZ.
Product Description
Clindamycin hydrochloride, European Pharmacopoeia (EP) Reference Standard
Supelco
Clindamycin hydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Clindamycin hydrochloride, lincosamide antibiotic
Sigma-Aldrich
Clarithromycin, 96.0-102.0% (HPLC)
Clarithromycin, European Pharmacopoeia (EP) Reference Standard
Clarithromycin for peak identification, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Clarithromycin, ≥95% (HPLC)
Supelco
Clarithromycin, Pharmaceutical Secondary Standard; Certified Reference Material