Merck
  • Removal of cations using ion-binding terpolymer involving 2-amino-6-nitro-benzothiazole and thiosemicarbazide with formaldehyde by batch equilibrium technique.
Removal of cations using ion-binding terpolymer involving 2-amino-6-nitro-benzothiazole and thiosemicarbazide with formaldehyde by batch equilibrium technique.
Journal of hazardous materials (2013-01-29)
Mohamed A Riswan Ahamed, Duraisamy Jeyakumar, Abdul R Burkanudeen
ABSTRACT
2-Amino-6-nitro-benzothiazole and thiosemicarbazide with formaldehyde (BTF) terpolymer was synthesized by the condensation polymerization technique. The elemental analysis and physico-chemical parameters of the terpolymer were measured. This chelation terpolymer was characterized by infrared, electronic and nuclear magnetic resonance ((1)H &(13)C NMR) spectral studies. The molecular weight of the terpolymer was determined by gel permeation chromatography (GPC). Surface analysis of the terpolymer was analyzed by scanning electron microscopy (SEM) and X-ray diffraction (XRD) method. The thermal stability of the terpolymer was analyzed by thermogravimetric analysis (TGA). The cation-exchange property of the terpolymer was determined by batch equilibrium method with the effect of pH, contact time and electrolytes. The reusability of the resin was also studied to estimate the effectiveness of the terpolymer resin.
Sigma-Aldrich
Thiosemicarbazide, puriss. p.a., 98%
Sigma-Aldrich
Thiosemicarbazide, 99%