Merck
  • Application of scaffolds for bone regeneration strategies: current trends and future directions.
Application of scaffolds for bone regeneration strategies: current trends and future directions.
Injury (2013-02-13)
Gry Hulsart Billström, Ashley W Blom, Sune Larsson, Andrew D Beswick
ABSTRACT
Scaffolds are extensively used in surgery to replace missing bone and to achieve bony union and fusion. An ideal scaffold should not only maintain, induce, and restore biological functions where cells, extracellular matrix, and growth factors are needed, but also have the right properties with respect to degradation, cell binding, cellular uptake, non-immunogenicity, mechanical strength, and flexibility. Here we examine both the basic science behind the development of scaffolds and comprehensively and systematically review the clinical applications.
Sigma-Aldrich
Lithium chloride, powder, ≥99.98% trace metals basis
Supelco
Lithium chloride solution, 1 M in ethanol
Sigma-Aldrich
Strontium, granular, 99% trace metals basis
Sigma-Aldrich
Strontium, dendritic pieces, purified by distillation, 99.99% trace metals basis
Sigma-Aldrich
Lithium chloride, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Sigma-Aldrich
Lithium chloride, AnhydroBeads, −10 mesh, 99.998% trace metals basis
Sigma-Aldrich
Strontium, random pieces, 99%
Sigma-Aldrich
Strontium, dendritic pieces, purified by distillation, 99.9% trace metals basis
Sigma-Aldrich
Lithium chloride, BioUltra, for molecular biology, anhydrous, ≥99.0% (AT)
Sigma-Aldrich
Lithium chloride, BioXtra, ≥99.0% (titration)
Sigma-Aldrich
Lithium chloride, for molecular biology, ≥99%
Sigma-Aldrich
Lithium chloride solution, 8 M, for molecular biology, ≥99%
Sigma-Aldrich
Lithium chloride, ReagentPlus®, 99%
Sigma-Aldrich
Lithium chloride, puriss. p.a., anhydrous, ≥99.0% (AT)
Sigma-Aldrich
Lithium chloride, ACS reagent, ≥99%