Merck
  • A bowl-shaped organic host using bispyridine ligands: selective encapsulation of carbonyl guests in water.
A bowl-shaped organic host using bispyridine ligands: selective encapsulation of carbonyl guests in water.
Chemical communications (Cambridge, England) (2013-01-23)
Kohei Yazaki, Norifumi Kishi, Munetaka Akita, Michito Yoshizawa
ABSTRACT
A bowl-shaped organic host was prepared by linking two anthracene-embedded bispyridine ligands with two methylene spacers. The water-soluble host has a hemispherical hydrophobic cavity (∼1 nm in diameter) with two cationic methylenebispyridinyl (Lewis acidic) moieties and shows the selective recognition and encapsulation of aromatic guest molecules containing carbonyl groups in water.
Sigma-Aldrich
Anthracene, sublimed grade, ≥99%
Supelco
Anthracene, analytical standard
Supelco
Anthracene, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Anthracene, reagent grade, 97%
Sigma-Aldrich
Anthracene, ReagentPlus®, 99%
Supelco
Anthracene, analytical standard
Sigma-Aldrich
Anthracene, suitable for scintillation, ≥99.0% (GC)