Merck
  • Cyclodextrin-crosslinked poly(acrylic acid): Synthesis, physicochemical characterization and controlled release of diflunisal and fluconazole from hydrogels.
Cyclodextrin-crosslinked poly(acrylic acid): Synthesis, physicochemical characterization and controlled release of diflunisal and fluconazole from hydrogels.
International journal of pharmaceutics (2013-01-19)
Marguerite J Kutyła, Lynette K Lambert, Nigel M Davies, Ross P McGeary, P Nicholas Shaw, Benjamin P Ross
ABSTRACT
The aim of this work was to develop mucoadhesive hydrogels with variable drug delivery properties by crosslinking poly(acrylic acid) (PAA) with cyclodextrins (CDs). CD-PAA polymers with high CD content and good inter-batch reproducibility were synthesized by activating PAA with SOCl2, then reacting PAA chloride with CD in the presence of 4-dimethylaminopyridine at 50°C. Manipulation of the synthesis conditions affected the physicochemical character of the CD-PAA polymers and hydrogels in terms of CD content, the average number of ester bonds to an individual CD, viscosity, and the association and release of model drugs. Inclusion complexation of diflunisal (DIF) and fluconazole (FLZ) with CD-PAA hydrogels was assessed by (19)F NMR spectroscopy and association constants (Kas) for DIF were in the range 220-486M(-1) with βCD-PAA and 1327-6055M(-1) with hydroxypropyl-βCD-PAA. For FLZ the Ka range was 34-171M(-1) with hydroxypropyl-βCD-PAA. The hydrogels were found to release both drugs by means of Fickian diffusion as the predominant mechanism. A slight trend toward negative correlation was found between the Ka and Higuchi kH values for DIF. These results highlight the potential of CD-PAA hydrogels to control the release of model drugs through inclusion complexation.
Sigma-Aldrich
Poly(acrylic acid, sodium salt) solution, average Mw ~1,200, 45 wt. % in H2O
Sigma-Aldrich
Poly(acrylic acid) solution, average Mw ~250,000, 35 wt. % in H2O
Sigma-Aldrich
Poly(acrylic acid) solution, average Mw ~100,000, 35 wt. % in H2O
Sigma-Aldrich
Poly(acrylic acid) solution, average Mw ~2,000, 50 wt. % in H2O, electronic grade
Sigma-Aldrich
Poly(acrylic acid, sodium salt) solution, average Mw ~8,000, 45 wt. % in H2O
Supelco
Diflunisal, analytical standard
Sigma-Aldrich
Poly(acrylic acid), average Mv ~4,000,000
Sigma-Aldrich
Poly(acrylic acid)
Sigma-Aldrich
Poly(acrylic acid sodium salt), average Mw ~2,100
Sigma-Aldrich
Poly(acrylic acid), average Mv ~3,000,000
Sigma-Aldrich
Poly(acrylic acid), average Mv ~1,250,000
Supelco
Poly(acrylic acid), analytical standard, average Mn 130,000 (Typical)
Sigma-Aldrich
Poly(acrylic acid), average Mv ~450,000
Sigma-Aldrich
Poly(acrylic acid sodium salt), average Mw ~5,100 by GPC, powder
Supelco
Poly(acrylic acid sodium salt), analytical standard, for GPC, 16,000