Merck
  • Effect of nonionic surfactant Brij 35 on the fate and transport of oxytetracycline antibiotic in soil.
Effect of nonionic surfactant Brij 35 on the fate and transport of oxytetracycline antibiotic in soil.
Journal of environmental management (2013-01-09)
Eman M Elsayed, Shiv O Prasher, Ramanbhai M Patel
ABSTRACT
In many parts of the world, river water is used for irrigation. Treated, partially treated, and even untreated water from wastewater treatment plants is discharged directly into rivers, thereby degrading the quality of the water. Consequently, irrigation water may contain surfactants which may affect the fate and transport of chemicals such as pesticides and antibiotics in agricultural soils. A field lysimeter study was undertaken to investigate the effect of the nonionic surfactant, Brij 35, on the fate and transport of an antibiotic, Oxytetracycline, commonly used in cattle farms. Nine PVC lysimeters, 1.0 m long × 0.45 m diameter, were packed with a sandy soil to a bulk density of 1.35 Mg m(-3). Cattle manure, containing Oxytetracycline, was applied at the surface of the lysimeters at the recommended rate of 10 t/ha. Each of three aqueous Brij 35 solutions, 0, 0.5 and 5 g L(-1) (i.e., 'good,' 'poor' and 'very poor' quality irrigation water) were each applied to the lysimeters in triplicate. Over a 90 day period, soil and leachate samples were collected and analyzed. Batch experiment results showed that the presence of the nonionic surfactant Brij 35 significantly reduced the sorption coefficient of OTC from 23.55 mL g(-1) in the aqueous medium to 19.49, 12.49 and 14.53 in the presence of Brij 35 at concentrations of 0.25, 2.5 and 5 g L(-1), respectively. Lysimeter results indicted the significant downward movement of OTC at depths of 60 cm into soil profile and leachate in the presence of surfactant. Thus, the reuse of wastewater containing surfactants might enhance the mobility of contaminants and increase ground water pollution.
Product Description
Oxytetracycline hydrochloride, European Pharmacopoeia (EP) Reference Standard
Supelco
Oxytetracycline hydrochloride, VETRANAL®, analytical standard
Sigma-Aldrich
Oxytetracycline hydrochloride
Sigma-Aldrich
Oxytetracycline dihydrate, Assay (anhydrous basis) 95.0%-102.0%
Millipore
Oxytetra Selective Supplement, suitable for microbiology